Skip to Content
Merck
All Photos(1)

Documents

79710

Sigma-Aldrich

O-Phospho-DL-serine

≥98.0% (NT), suitable for cell culture

Synonym(s):

DL-2-Amino-3-hydroxypropanoic acid 3-phosphate, DL-SOP, DL-Serine monophosphoric acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H8NO6P
CAS Number:
Molecular Weight:
185.07
Beilstein:
1726828
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

product name

O-Phospho-DL-serine, ≥98.0% (NT)

Assay

≥98.0% (NT)

form

powder or crystals

technique(s)

cell culture | mammalian: suitable

impurities

≤0.5% free serine (TLC)
≤1% water

color

white to faint beige

SMILES string

NC(COP(O)(O)=O)C(O)=O

InChI

1S/C3H8NO6P/c4-2(3(5)6)1-10-11(7,8)9/h2H,1,4H2,(H,5,6)(H2,7,8,9)

InChI key

BZQFBWGGLXLEPQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

O-Phospho-DL-serine (pSer) is used in the study of non-enzymatic aminophospholipid glycation.
O-Phosphoserine is used in alternative pathways for biosynthesis of cysteine and of selenocysteine (Sec) by tRNA-dependent conversion of O-phosphoserine (Sep) by O-phosphoseryl-tRNA:selenocysteinyl-tRNA synthase (SepSecS).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuhei Araiso et al.
Nucleic acids research, 36(4), 1187-1199 (2007-12-26)
The micronutrient selenium is present in proteins as selenocysteine (Sec). In eukaryotes and archaea, Sec is formed in a tRNA-dependent conversion of O-phosphoserine (Sep) by O-phosphoseryl-tRNA:selenocysteinyl-tRNA synthase (SepSecS). Here, we present the crystal structure of Methanococcus maripaludis SepSecS complexed with
Yoko Chiba et al.
The Journal of biological chemistry, 288(16), 11448-11458 (2013-03-13)
Novel-type serine-synthesizing enzymes, termed metal-independent phosphoserine phosphatases (iPSPs), were recently identified and characterized from Hydrogenobacter thermophilus, a chemolithoautotrophic bacterium belonging to the order Aquificales. iPSPs are cofactor-dependent phosphoglycerate mutase (dPGM)-like phosphatases that have significant amino acid sequence similarity to dPGMs
Pengcheng Wang et al.
Plant physiology, 161(3), 1392-1408 (2013-01-10)
Reactive oxygen species (ROS) have been characterized as both important signaling molecules and universal stressors that mediate many developmental and physiological responses. So far, details of the transcriptional mechanism of ROS-responsive genes are largely unknown. In the study reported here
Daniel Agren et al.
The Journal of biological chemistry, 283(46), 31567-31574 (2008-09-19)
The biosynthesis of cysteine is a crucial metabolic pathway supplying a building block for de novo protein synthesis but also a reduced thiol as a component of the oxidative defense mechanisms that appear particularly vital in the dormant state of
Po-Hsiang Wang et al.
The ISME journal, 13(4), 1042-1055 (2019-01-05)
Microbes in ecosystems often develop coordinated metabolic interactions. Therefore, understanding metabolic interdependencies between microbes is critical to deciphering ecosystem function. In this study, we sought to deconstruct metabolic interdependencies in organohalide-respiring consortium ACT-3 containing Dehalobacter restrictus using a combination of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service