Skip to Content
Merck

Skip To

78551

3-(4-Methylbenzylidene)camphor

≥98.0% (GC)

Synonym(s):

1,7,7-Trimethyl-3-(4-methylbenzylidene)-norbornan-2-one, 1,7,7-Trimethyl-3-[(4-methylphenyl)methylene]-bicyclo[2.2.1]heptan-2-one, 4-MBC, 4-Methylbenzylidene camphor

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C18H22O
CAS Number:
Molecular Weight:
254.37
PubChem Substance ID:
UNSPSC Code:
12352212
MDL number:
NACRES:
NA.25
EC Number:
253-242-6
Beilstein/REAXYS Number:
9213949
Assay:
≥98.0% (GC)
Form:
crystals
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98.0% (GC)

form

crystals

SMILES string

Cc1ccc(cc1)\C=C2\[C@@H]3CC[C@](C)(C2=O)C3(C)C

InChI

1S/C18H22O/c1-12-5-7-13(8-6-12)11-14-15-9-10-18(4,16(14)19)17(15,2)3/h5-8,11,15H,9-10H2,1-4H3/b14-11-/t15-,18+/m0/s1

InChI key

HEOCBCNFKCOKBX-IRELMECMSA-N

General description

An estrogenic endocrine disruptor found to affect the development processes of ductal outgrowth and branching morphogenesis.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
61547PHR28051424222
form

crystals

form

-

form

solid

form

-

assay

≥98.0% (GC)

assay

-

assay

-

assay

-

Quality Level

100

Quality Level

300

Quality Level

300

Quality Level

-


Still not finding the right product?

Explore all of our products under 3-(4-Methylbenzylidene)camphor


pictograms

Environment

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

372.2 °F

flash_point_c

189 °C

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - ED HH 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters