Skip to Content

77631

(R)-Mevalonic acid 5-pyrophosphate tetralithium salt

≥95% (TLC)

Synonym(s):

(R)-Mevalonic acid 5-diphosphate tetralithium salt, R-MVAPP

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C6H10Li4O10P2
CAS Number:
Molecular Weight:
331.85
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352106
MDL number:
Assay:
≥95% (TLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (TLC)

form

powder

optical activity

[α]/D -2.9±0.5°, c = 1 in hydrochloric acid (0.02 M)

storage temp.

−20°C

SMILES string

[Li+].[Li+].[Li+].[Li+].C[C@@](O)(CCOP([O-])(=O)OP([O-])([O-])=O)CC([O-])=O

InChI

1S/C6H14O10P2.4Li/c1-6(9,4-5(7)8)2-3-15-18(13,14)16-17(10,11)12;;;;/h9H,2-4H2,1H3,(H,7,8)(H,13,14)(H2,10,11,12);;;;/q;4*+1/p-4/t6-;;;;/m1..../s1

InChI key

GFIXUUFXTAPBAD-CZNRRTBNSA-J

Biochem/physiol Actions

Metabolite of the mevalonate pathway, which plays a key role in the biosynthesis of sterols, dolichol, heme and ubiquinone. Of interest for research in the disease areas oncology, autoimmune diseases, artherosclerosis and Alzheimer disease, as well as for inherited deficiencies of mevalonate kinase.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library