Skip to Content

76200

D-Panthenol

≥98.0% (NT)

Synonym(s):

(R)-(+)-2,4-Dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutyramide, (R)-2,4-Dihydroxy-3,3-dimethylbutyric 3-hydroxypropylamide, D-Pantothenyl alcohol, Dexpanthenol, Provitamin B

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C9H19NO4
CAS Number:
Molecular Weight:
205.25
UNSPSC Code:
12352104
NACRES:
NA.28
PubChem Substance ID:
EC Number:
201-327-3
Beilstein/REAXYS Number:
1724947
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic

Quality Segment

assay

≥98.0% (NT)

form

liquid

optical activity

[α]20/D +30±1°, c = 5% in H2O

impurities

≤0.002% heavy metals, ≤0.5% 3-aminopropanol

ign. residue

≤0.1% (as SO4)

color

colorless

refractive index

n20/D 1.496-1.500

density

1.20 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)(CO)[C@@H](O)C(=O)NCCCO

InChI

1S/C9H19NO4/c1-9(2,6-12)7(13)8(14)10-4-3-5-11/h7,11-13H,3-6H2,1-2H3,(H,10,14)/t7-/m0/s1

InChI key

SNPLKNRPJHDVJA-ZETCQYMHSA-N

Application


  • The combination of allantoin, bisabolol, D-panthenol and dipotassium glycyrrhizinate mitigates UVB-induced PGE(2) synthesis by keratinocytes.: This study investigates the protective effects of D-panthenol, combined with other compounds, against UVB-induced inflammation in keratinocytes. The results highlight D-panthenol′s role in skin care formulations aimed at reducing UV-induced damage and inflammation (Tan et al., 2024).

  • Protective Effect of D-Panthenol in Rhabdomyolysis-Induced Acute Kidney Injury.: This research explores the protective properties of D-panthenol in mitigating acute kidney injury caused by rhabdomyolysis. The findings suggest potential therapeutic applications for D-panthenol in renal protection and recovery (Semenovich et al., 2022).

  • Alpha-Glycerylphosphorylcholine and D-Panthenol Eye Drops in Patients Undergoing Cataract Surgery.: This clinical study examines the efficacy of eye drops containing D-panthenol in patients undergoing cataract surgery, demonstrating its potential to enhance postoperative recovery and eye health (Martone et al., 2022).

  • Common Cosmetic Compounds Can Reduce Air Pollution-Induced Oxidative Stress and Pro-Inflammatory Response in the Skin.: This study highlights the role of D-panthenol in protecting the skin against oxidative stress and inflammation caused by air pollution, emphasizing its importance in cosmetic formulations for urban environments (Hergesell et al., 2022).

Biochem/physiol Actions

D-Panthenol is the alcohol analogue and biological precursor of D-pantothenic acid. These analogues are precursors in the biosynthesis of coenzyme A. D-Panthenol and D-Pantothenic acid are used in a variety of skin protection products. D-pantothenic acid may have a role in controlling keratinocyte proliferation and differentiation. D-Panthenol may be used for studies of skin protection emulsions from UV irradiation and as a humectant.


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library