Skip to Content
Merck
All Photos(1)

Key Documents

76157

Sigma-Aldrich

(4R)-4-Hydroxy-L-glutamic acid

≥98.0% (TLC)

Synonym(s):

erythro-(4R)-4-Hydroxy-L-glutamic acid, H-(2S,4R)-γ-Hydroxy-Glu-OH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9NO5
CAS Number:
Molecular Weight:
163.13
Beilstein:
1725871
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.28

product name

(4R)-4-Hydroxy-L-glutamic acid, ≥98.0% (TLC)

Quality Level

Assay

≥98.0% (TLC)

form

powder

optical activity

[α]/D 20.5±1.5°, c = 1 in H2O

color

white

mp

171 °C

storage temp.

−20°C

SMILES string

N[C@@H](C[C@@H](O)C(O)=O)C(O)=O

InChI

1S/C5H9NO5/c6-2(4(8)9)1-3(7)5(10)11/h2-3,7H,1,6H2,(H,8,9)(H,10,11)/t2-,3+/m0/s1

InChI key

HBDWQSHEVMSFGY-STHAYSLISA-N

Biochem/physiol Actions

(4R)-4-Hydroxy-L-glutamic acid or (2S,4R)-4-hydroxyglutamate was shown to activate the metabotropic glutamate receptors, mGlu1a, mGlu2, and mGlu8a in a dose-dependent manner.
Substrate for aminotransferase; pharmacological characterization at human glutamate transporter subtypes 1-3; for studying structure-activity relationships (SAR) for ionotropic and metabotropic glutamate receptors.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A S Bessis et al.
Bioorganic & medicinal chemistry letters, 11(12), 1569-1572 (2001-06-20)
The (2S,4R)- and (2S,4S)-4-hydroxyglutamates activate cloned mGlu(1a), mGlu(2), and mGlu(8a) receptors with different potencies. Best results were obtained with the (2S,4S) isomer being almost as potent as glutamate on mGlu(1a)R and mGlu(8a)R. Data are interpreted on the basis of the
Metabolism of gamma-hydroxyglutamic acid. I. Conversion to alpha-hydroxy-gamma-ketoglutarate by purified glutamic-aspartic transaminase to rat liver.
A GOLDSTONE et al.
The Journal of biological chemistry, 237, 3476-3485 (1962-11-01)
Lennart Bunch et al.
ChemMedChem, 4(11), 1925-1929 (2009-09-05)
Subtype-selective ligands are of great interest to the scientific community, as they provide a tool for investigating the function of one receptor or transporter subtype when functioning in its native environment. Several 4-substituted (S)-glutamate (Glu) analogues were synthesized, and altogether
Sebastien Alaux et al.
Journal of medicinal chemistry, 48(25), 7980-7992 (2005-12-13)
A series of nine L-2,4-syn-4-alkylglutamic acid analogues (1a-i) were synthesized in high yield and high enantiomeric excess (>99% ee) from their corresponding 4-substituted ketoglutaric acids (2a-i), using the enzyme aspartate aminotransferase (AAT) from pig heart or E. coli. The synthesized

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service