Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.32
UNSPSC Code:
12352116
product line
BioReagent
Quality Segment
assay
≥90% (HPLC)
form
solid
mol wt
Mw 858 g/mol
manufacturer/tradename
ATTO-TEC GmbH
λ
in methanol: water (1:1) (with 0.1% perchloric acid)
UV absorption
λ: 502-508 nm Amax
suitability
suitable for fluorescence
storage temp.
−20°C
General description
Atto 488 is a fluorescent label with excellent water solubility. Characteristic features of the label are strong absorption, high fluorescence quantum yield, high photostability, and very little triplet formation. Thus Atto 488 is highly suitable for single-molecule detection applications and high-resolution microscopy such as PALM, dSTORM, STED etc. Additionally the dye highly qualifies to be applied in flow cytometry (FACS), fluorescence in-situ hybridization (FISH) and many more. The fluorescence is excited most efficiently in the range 480 - 515 nm. For instance the 488 nm line of the Argon-Ion laser is very suitable for excitation.
find more information here
find more information here
Legal Information
This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Analysis of fluorescent nanostructures in biological systems by means of spectral position determination microscopy (SPDM).
Muller, P., et al. et al.
Current Microscopy Contributions to Advances in Science and Technology, 1, 3-12 (2012)
Monitoring single membrane protein dynamics in a liposome manipulated in solution by the ABELtrap.
Rendler, T., et al.
arXiv, 1102-1102 (2011)
Mohd A Mohd Ridzuan et al.
PloS one, 7(3), e33845-e33845 (2012-04-06)
An actomyosin motor complex assembled below the parasite's plasma membrane drives erythrocyte invasion by Plasmodium falciparum merozoites. The complex is comprised of several proteins including myosin (MyoA), myosin tail domain interacting protein (MTIP) and glideosome associated proteins (GAP) 45 and
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 74417-1MG | 04061826688120 |