Skip to Content
Merck

70718

Methoxypolyethylene glycol 5,000 acetic acid

≥80%

Synonym(s):

Polyethylene glycol, mono-Methyl polyethylene glycol 5,000 acetic acid, O-(Carboxymethyl)-O′-methylpolyethylene glycol 5′000

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352106
NACRES:
NA.25
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥80%

form

powder

Ω-end

carboxylic acid

α-end

methoxy

storage temp.

−20°C



Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library