Skip to Content
Merck
All Photos(1)

Documents

70124

Sigma-Aldrich

4-Nitrophenyl myristate

lipase substrate, ≥95.0% (HPLC), powder

Synonym(s):

4-Nitrophenyl tetradecanoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H31NO4
CAS Number:
Molecular Weight:
349.46
Beilstein:
2004374
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

product name

4-Nitrophenyl myristate, ≥95.0% (HPLC)

Quality Level

Assay

≥95.0% (HPLC)

form

powder

solubility

chloroform: 50 mg/mL, clear to very slightly hazy, colorless to dark yellow

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCC(=O)Oc1ccc(cc1)[N+]([O-])=O

InChI

1S/C20H31NO4/c1-2-3-4-5-6-7-8-9-10-11-12-13-20(22)25-19-16-14-18(15-17-19)21(23)24/h14-17H,2-13H2,1H3

InChI key

ZBBNFJIVAHGZKA-UHFFFAOYSA-N

Application

4-Nitrophenyl myristate has been used as a substrate to measure the activity of:
  • bile salt-activated lipase
  • esterase
  • neutral lipase

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qiaoqiao Sun et al.
Journal of bioscience and bioengineering, 122(5), 539-544 (2016-10-26)
Thermophilic Neosartorya fischeri P1 is an excellent lipase producer and harbors seven lipase genes. All genes were found to be functional after heterologous expression in Escherichia coli. One of them, LIP09, showed high-level expression in Pichia pastoris with the yield
Mirella Vazzana et al.
The Journal of the Acoustical Society of America, 147(4), 2414-2414 (2020-05-04)
Underwater sounds generated by anthropogenic activity can cause behavior changes, temporary loss of hearing, damage to parts of the body, or death in a number of marine organisms and can also affect healing and survival. In this study, the authors
Vahid Morshedi et al.
Comparative biochemistry and physiology. Part A, Molecular & integrative physiology, 261, 111044-111044 (2021-08-10)
The present research was conducted to provide insight into digestive larval capacity in Acanthopagrus latus larvae from hatching up to 30 days after hatching (DAH). Newly hatched larvae were stocked into six 300-L cylindrical polyethylene tanks at a density of larvae
Maria Stella Cappello et al.
Systematic and applied microbiology, 33(8), 461-467 (2010-11-26)
The diversity of indigenous Oenococcus oeni strains was investigated by molecular and biochemical characterization of isolates from Malvasia Nera wine, an economically important red wine of the Salento Region (Apulia, Italy), during spontaneous malolactic fermentation (MLF). A total of 82
Effects of increasing protein level on the performance, enzyme activity and body composition of the Brazilian sardine, Sardinella brasiliensis (Steindachner, 1879)
Sterzelecki FC, et al.
Aquaculture Nutrition, 366-374 null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service