Skip to Content
Merck

Skip To

69658

Fluorescein isothiocyanate–dextran

average mol wt 150,000

Synonym(s):

FITC–Dextran 150, FITC–Dextran

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352201
NACRES:
NA.32
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic

Quality Segment

conjugate

FITC conjugate

form

powder or crystals

mol wt

average mol wt 150,000

extent of labeling

0.001-0.020 mol/mol FITC glucose

fluorescence

λex 493 nm; λem 520 nm±5 nm in 0.1 M Tris pH 8.0

application(s)

cell analysis

Application

Dextran labeled with fluorescein isothiocyanate for possible use in perfusion studies in animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
524714694590718
description

average mol wt 150,000

description

average mol wt 2,000,000

description

average mol wt 70,000, (FITC:Glucose = 1:250)

description

average Mw 70,000

biological source

synthetic

biological source

synthetic

biological source

synthetic

biological source

synthetic

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

form

powder or crystals

form

powder or crystals

form

powder

form

powder or crystals

extent of labeling

0.001-0.020 mol/mol FITC glucose

extent of labeling

0.001-0.020 mol FITC per mol glucose (substitution)

extent of labeling

~0.004 mol/mol FITC glucose

extent of labeling

0.001-0.020 mol/mol FITC glucose

mol wt

average mol wt 150,000

mol wt

average mol wt 2,000,000

mol wt

average mol wt 70,000

mol wt

average Mw 70,000

conjugate

FITC conjugate

conjugate

FITC conjugate

conjugate

FITC conjugate

conjugate

FITC conjugate


Still not finding the right product?


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters