Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C18H22N6O12P2 · xC6H15N
CAS Number:
Molecular Weight:
576.35 (free acid basis)
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.32
Quality Segment
assay
≥95.0% (HPLC)
form
liquid
pH
7.0-8.0
storage temp.
−20°C
SMILES string
OP(O)(OP(O)(OC[C@@H]1[C@@H]([R1])[C@@H]([R2])[C@H](N(C=N2)C3=C2C(NC(N)=N3)=O)O1)=O)=O.CCN(CC)CC.O=C(C4=C(NC)C=CC=C4)O.O=C(C5=C(NC)C=CC=C5)O
Storage Class
10 - Combustible liquids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 69244-1.5UMOL | 04061826643587 |