Skip to Content
Merck
All Photos(1)

Key Documents

68120

Sigma-Aldrich

Malvidin chloride

≥95.0% (HPLC)

Synonym(s):

3,4′,5,7-Tetrahydroxy-3′,5′-dimethoxyflavylium chloride, Malvidin, Oenidin chloride, Primulidin chloride, Syringidin chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H15ClO7
CAS Number:
Molecular Weight:
366.75
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95.0% (HPLC)

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

SMILES string

[Cl-].COc1cc(cc(OC)c1O)-c2[o+]c3cc(O)cc(O)c3cc2O

InChI

1S/C17H14O7.ClH/c1-22-14-3-8(4-15(23-2)16(14)21)17-12(20)7-10-11(19)5-9(18)6-13(10)24-17;/h3-7H,1-2H3,(H3-,18,19,20,21);1H

InChI key

KQIKOUUKQBTQBE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Malvidin is a O-methylated anthocyanidin, a biologically active plant pigment.

Application

Malvidin chloride has been used as an inhibitor to screen its activity against electric eels acetylcholinesterase (AChE). It may have been used as an antioxidant to test its antiproliferation effects on Caco-2 colon and HepG2 liver cancer cells. Malvidin may be used as a reference standard in the analysis of anthocyanidins extracted from Kenyan purple tea extract using high-performance liquid chromatography (HPLC).

Biochem/physiol Actions

Anthocyanidin. Antioxidant.
Malvidin chloride is an excellent cell signaling inhibitor of 3′,5′-cyclic adenosine monophosphate (cAMP), phosphodiesterases (PDE), and certain cGMP-specific phosphodiesterase. It is also a known inhibitor of α-glucosidase.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ping-Hsiao Shih et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 43(10), 1557-1566 (2005-06-21)
Anthocyanins are naturally occurring reddish pigments that abundant in fruits and vegetables. To investigate the mechanistic basis for the anti-tumor properties of anthocyanins, five aglycone (cyanidin, delphinidin, malvidin, pelargonidin, and peonidin) and four glycosylated (cyanidin-3-glucoside, malvidin-3-glucoside, pelargonidin-3-glucoside and peonidin-3-glucoside) anthocyanins
Jin Won Hyun et al.
Journal of agricultural and food chemistry, 52(8), 2213-2217 (2004-04-15)
Oryza sativa cv. Heugjinjubyeo (Gramineae), anthocyanin-pigmented rice, having dark purple grains, is known broadly as enriched rice with an improved taste. Two bioactive compounds were isolated from the 0.5% HCl-ethyl alcohol soluble fraction of the aleurone layer of O. sativa
Mario Dell'Agli et al.
Journal of agricultural and food chemistry, 53(6), 1960-1965 (2005-03-17)
A moderate consumption of red wine may reduce the risk of cardiovascular diseases via wine-derived phenolic compounds. A variety of biological mechanisms have been proposed for wine-derived phenolic compounds including nitric oxide-mediated vasorelaxation. This study examined whether the vasodilating effect
Ellen Silva Lago-Vanzela et al.
Journal of agricultural and food chemistry, 59(24), 13136-13146 (2011-11-25)
The aim of this study was the detailed characterization of the phenolic composition and the determination of the antioxidant activity of the Bordô grape (Vitis labrusca) cultivated in South Brazil. The edible parts of Bordô grapes (flesh and skin) contained
V Amico et al.
Bioresource technology, 99(13), 5960-5966 (2007-12-07)
On the basis of a preliminary screening of seven different samples of Sicilian grape pomace, the 'Nerello Mascalese' sample NM2 was selected for an ethanol preparative extraction. The defatted NM2 EtOH extract was subjected to DPPH() and GAE assays, showing

Articles

Antioxidants protect biological systems from oxidative damage produced by oxygen-containing free radicals and from redoxactive transition metal ions such as iron, copper, and cadmium.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service