Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C15H18N2O3S · C2HF3O2
CAS Number:
Molecular Weight:
420.40
NACRES:
NA.32
PubChem Substance ID:
UNSPSC Code:
12352209
MDL number:
Quality Segment
assay
≥99.0% (sum of enantiomers, HPLC)
optical activity
[α]20/D +68±3°, c = 1% in methanol
storage temp.
2-8°C
SMILES string
OC(=O)C(F)(F)F.CSCC[C@H](N)C(=O)Nc1ccc2C(C)=CC(=O)Oc2c1
InChI
1S/C15H18N2O3S.C2HF3O2/c1-9-7-14(18)20-13-8-10(3-4-11(9)13)17-15(19)12(16)5-6-21-2;3-2(4,5)1(6)7/h3-4,7-8,12H,5-6,16H2,1-2H3,(H,17,19);(H,6,7)/t12-;/m0./s1
InChI key
VSAUWUJWXBUSEU-YDALLXLXSA-N
Application
L-Methionine 7-amido-4-methylcoumarin trifluoroacetate and its analogues may be used to study the kinetics of methionine aminopeptidase (hMetAP2). L-Methionine 7-amido-4-methylcoumarin trifluoroacetate may be use with other substrates to assess the specificity of various aminopeptidases.
Other Notes
Substrate for calpain
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 282650-500MG | 04061826228425 |
| 282650-1G | 04061826671443 |
| 64367-25MG | 04061833277768 |
