Skip to Content
Merck
All Photos(1)

Key Documents

63759

Sigma-Aldrich

7-Mercapto-4-methylcoumarin

≥97.0% (GC)

Synonym(s):

4-Methyl-7-thioumbelliferone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8O2S
CAS Number:
Molecular Weight:
192.23
Beilstein:
7476043
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥97.0% (GC)

form

solid

fluorescence

λex 358 nm; λem 385 nm in methylene chloride

SMILES string

CC1=CC(=O)Oc2cc(S)ccc12

InChI

1S/C10H8O2S/c1-6-4-10(11)12-9-5-7(13)2-3-8(6)9/h2-5,13H,1H3

InChI key

BYDNGJQDDNBAHI-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yizhi Zhang et al.
Small (Weinheim an der Bergstrasse, Germany), 14(20), e1704433-e1704433 (2018-04-18)
Isolating and in situ profiling the heterogeneous molecular phenotype of circulating tumor cells are of great significance for clinical cancer diagnosis and personalized therapy. Herein, an on-chip strategy is proposed that combines size-based microfluidic cell isolation with multiple spectrally orthogonal
Eleonora Cara et al.
Scientific reports, 8(1), 11305-11305 (2018-07-29)
Controlling the location and the distribution of hot spots is a crucial aspect in the fabrication of surface-enhanced Raman spectroscopy (SERS) substrates for bio-analytical applications. The choice of a suitable method to tailor the dimensions and the position of plasmonic
Yi-Hsuan Lee et al.
Scientific reports, 7, 39907-39907 (2017-01-07)
4-Hydroxybenzoate is a phenolic derivative of alkyl benzoates and is a widely used preservative in cosmetic and pharmaceutical products. The presence of 4-hydroxybenzoates in the human body may result from the use of pharmaceutical and personal care products. These compounds
Ying-Jung Wu et al.
Analytical and bioanalytical chemistry, 410(28), 7405-7414 (2018-09-08)
In the clinic, ethosuximide is commonly used to treat generalized absence seizures but has recently been repurposed for other diseases. Because of adverse effects and drug interactions, high-throughput therapeutic drug monitoring of ethosuximide is necessary. Microextraction is a simple, effective
Giacomo Dacarro et al.
Journal of colloid and interface science, 505, 1055-1064 (2017-07-13)
The synthesis of large pentatwinned five-branched gold nanostars (GNS) has been modified so to obtain overall dimensions shrunk to 60% and a lower branches aspect ratio, leading to a dramatic blue shift of their two near-infrared (NIR) localized surface plasmon

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service