Skip to Content
Merck

63226

Benzamidine hydrochloride 1 M solution

Synonym(s):

Benzamidine hydrochloride solution, Additive Screening Solution 37/Kit-No 78374

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
NACRES:
NA.26
PubChem Substance ID:
UNSPSC Code:
12352202
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


storage temp.

2-8°C

Quality Segment

SMILES string

Cl.NC(=N)c1ccccc1

InChI

1S/C7H8N2.ClH/c8-7(9)6-4-2-1-3-5-6;/h1-5H,(H3,8,9);1H

InChI key

LZCZIHQBSCVGRD-UHFFFAOYSA-N



Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library