62650
Lugol solution
for microscopy (Bact., Bot.)
Synonym(s):
Iodine/Potassium iodide solution
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
grade
for microscopy (Bact., Bot.)
technique(s)
microbe id | staining: suitable
density
1.007 g/mL at 20 °C
suitability
suitable for microscopy
protozoa
antibiotic activity spectrum
Gram-negative bacteria
Gram-positive bacteria
SMILES string
[K+].[I-].II
InChI
1S/I2.HI.K/c1-2;;/h;1H;/q;;+1/p-1
InChI key
XUEKKCJEQVGHQZ-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
General description
Lugol iodine solution is an effective preservative for the marine bacteria.
Lugol solution is a disinfectant, which is used to detect starch in organic compounds. It is used in dental procedures, diagnosis of cervical cell alterations and Schiller´s test. Lugol solution is used in panendoscopic examination of squamous cell carcinoma of the esophagus. It is used as a mordant in Gram staining.
Application
Lugol solution has been used as a staining component to detect starch in plants. It has also been used to saturate thyroid in biodistribution study.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
The Chemical Mechanism of Bacterial Behavior: I. Behavior Toward Dyes-Factors Controlling the Gram Reaction 1
Stearn EW and Stearn AE
Journal of Bacteriology, 9(5), 463-463 (1924)
Cryo-focused ion beam sample preparation for imaging vitreous cells by cryo-electron tomography
Schaffer M, et al.
Bio-protocol, 5.17, e1575-e1575 (2015)
A J Pomroy
Applied and environmental microbiology, 47(5), 1191-1192 (1984-05-01)
Lugol iodine solution was compared with glutaraldehyde as a preservative for marine bacteria. Direct counts with the fluorochrome 4',6-diamidino-2-phenylindole show no significant difference between the preservatives, but the use of Lugol solution has several advantages over glutaraldehyde, especially in the
Evaluation of the presence of arabinogalactan proteins and pectins during Quercus suber male gametogenesis
Costa M, et al.
Annals of Botany, 115(1), 81-92 (2014)
Lugol?s solution and other iodide preparations: perspectives and research directions in Graves? disease
Calissendorff J and Falhammar H
Endocrine, 58(3), 467-473 (2017)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service