Skip to Content
Merck

61028

Genapol® C-100

Synonym(s):

Coceth-10, Coconut oil alcohol, ethoxylated

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12161900
NACRES:
NA.25
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

non-ionic

mol wt

Mw 627

CMC

0.075

solubility

H2O: 1 g/10 mL, clear, colorless

Other Notes

Major component: Polyoxyethylene (10) dodecyl ether

Legal Information

Genapol is a registered trademark of Clariant GmbH


signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Dam. 1

Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

482.0 °F - open cup

flash_point_c

250 °C - open cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library