Skip to Content
Merck
All Photos(1)

Key Documents

54765

Sigma-Aldrich

γ-Carotene

≥90% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H56
CAS Number:
Molecular Weight:
536.87
Beilstein:
3226412
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.32

Assay

≥90% (HPLC)

form

solid

mol wt

536.87 g/mol

λ

in hexane (with 2% dichloromethane)

UV absorption

λ: 458-462 nm Amax

storage temp.

−20°C

SMILES string

CC(/C=C/C1=C(C)CCCC1(C)C)=C\C=C\C(C)=C\C=C\C=C(C)\C=C\C=C(\C=C\C=C(C)\CCC=C(C)C)C

InChI

1S/C40H56/c1-32(2)18-13-21-35(5)24-15-26-36(6)25-14-22-33(3)19-11-12-20-34(4)23-16-27-37(7)29-30-39-38(8)28-17-31-40(39,9)10/h11-12,14-16,18-20,22-27,29-30H,13,17,21,28,31H2,1-10H3/b12-11+,22-14+,23-16+,26-15+,30-29+,33-19+,34-20+,35-24+,36-25+,37-27+

InChI key

HRQKOYFGHJYEFS-BXOLYSJBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Research area: Cell Signaling

Carotenoids are tetraterpene pigments known for displaying a range of colors such as yellow, orange, red, and purple. They are mostly found in photosynthetic bacteria, certain species of archaea and fungi, algae, plants, and animals. γ-Carotene, characterized by its unsubstituted β-ionone rings, serves as a precursor to retinoids and is referred to as pro-vitamin A.

Biochem/physiol Actions

Metabolite in carotenoid biosynthesis and the biosynthesis of plant secondary metabolites.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takashi Maoka et al.
Journal of agricultural and food chemistry, 63(5), 1622-1630 (2015-01-31)
Three new oxidative metabolites of lycopenes, (erythro)-lycopene-5,6-diol, (threo)-lycopene-5,6-diol, and 1,16-dehydro-2,6-cyclolycopene-5-ol B, and four new oxidative metabolites of γ-carotenes, 2',6'-cyclo-γ-carotene-1',5'-diol A, 2',6'-cyclo-γ-carotene-1',5'-diol B, (erythro)-γ-carotene-5,6-diol, and (threo)-γ-carotene-5,6-diol, were isolated as minor components from the aril of gac, Momordica cochinchinensis. These structures were
C Schmidt-Dannert et al.
Nature biotechnology, 18(7), 750-753 (2000-07-11)
The burgeoning demand for complex, biologically active molecules for medicine, materials science, consumer products, and agrochemicals is driving efforts to engineer new biosynthetic pathways into microorganisms and plants. We have applied principles of breeding, including mixing genes and modifying catalytic
Triantafyllos Roukas et al.
Applied biochemistry and biotechnology, 175(1), 182-193 (2014-09-25)
The effect of the aeration rate on carotene production from deproteinized hydrolyzed whey by Blakeslea trispora in a bubble column reactor was investigated. Aeration rate significantly affected carotene concentration and morphology of the fungus. Enhanced aeration caused change of the
P. D. Fraser et al.
Plant physiology, 105(1), 405-413 (1994-05-01)
Tomato (Lycopersicon esculentum Mill. cv Ailsa Craig) fruit, at five stages of development, have been analyzed for their carotenoid and chlorophyll (Chl) contents, in vitro activities of phytoene synthase, phytoene desaturase, and lycopene cyclase, as well as expression of the
Carotenoids as natural functional pigments
Maoka T
Journal of Natural Medicines, 74(1), 1?16-1?16 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service