Skip to Content
Merck
All Photos(1)

Documents

53727

Sigma-Aldrich

N-(3-Hydroxydodecanoyl)-DL-homoserine lactone

≥97% (HPLC)

Synonym(s):

3-Hydroxy-N-(tetrahydro-2-oxo-3-furanyl)dodecanamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H29NO4
CAS Number:
Molecular Weight:
299.41
MDL number:
UNSPSC Code:
41116134
PubChem Substance ID:
NACRES:
NA.25

Assay

≥97% (HPLC)

composition

carbon, 63.5-64.9%
nitrogen, 4.4-4.9%

storage temp.

−20°C

SMILES string

CCCCCCCCCC(O)CC(=O)NC1CCOC1=O

InChI

1S/C16H29NO4/c1-2-3-4-5-6-7-8-9-13(18)12-15(19)17-14-10-11-21-16(14)20/h13-14,18H,2-12H2,1H3,(H,17,19)

InChI key

RGTXFFYJJNWEPV-UHFFFAOYSA-N

Biochem/physiol Actions

N-(3-Hydroxydodecanoyl)-DL-homoserine lactone is a member of N-acyl-homoserine lactone family. N-Acylhomoserine lactones (AHL) regulate gene expression in gram-negative bacteria, such as Echerichia and Salmonella, and are involved in quorum sensing, cell to cell communication among bacteria; for reviews see. Bacterial intercellular communication has become a target for the development of new anti-virulence drugs, and a research focus for the prevention of biofilm formation.
Quorum-sensing signal generation

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Passador et al.
Journal of bacteriology, 178(20), 5995-6000 (1996-10-01)
A series of structural analogs of the Pseudomonas aeruginosa autoinducer [PAI, N-3-oxo-dodecanoyl homoserine lactone] were obtained and tested for their ability to act as autoinducers in stimulating the expression of the gene for elastase (lasB) by measuring beta-galactosidase production from
B M Davis et al.
Biophysical journal, 101(1), 245-254 (2011-07-05)
A practical label-free method for the rapid determination of small-molecule critical micelle concentration (CMC) using a fixed-angle light-scattering technique is described. Change in 90° light scattering at a fixed wavelength of incident radiation with increasing bacterial quorum molecule concentration and
Tommaso R I Cataldi et al.
Journal of mass spectrometry : JMS, 43(1), 82-96 (2007-08-22)
A method for the comprehensive profiling of the N-acyl-homoserine lactone (AHL) family of bacterial quorum-sensing molecules is presented using liquid chromatography (LC) coupled to a hybrid quadrupole linear ion trap (LTQ) and Fourier-transform ion-cyclotron-resonance mass spectrometer (FTICR). We demonstrate an
Kaoru Yamabe et al.
Biomedical research (Tokyo, Japan), 41(6), 259-268 (2020-12-04)
Recently, opportunistic nosocomial infections caused by Acinetobacter baumannii have become increasingly prevalent worldwide. The pathogen often establishes biofilms that adhere to medical devices, causing chronic infections refractory to antimicrobial therapy. Clinical reports have indicated that some macrolide antibiotics are effective
Meng Gui et al.
Frontiers in microbiology, 9, 1712-1712 (2018-08-16)
Sturgeon is an important fresh water-culture fish in China. A problem with sturgeon is its high susceptibility to spoilage. Food spoilage is reported to be regulated by quorum sensing (QS). To identify the QS signals acetylated homoserine lactones (AHLs) in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service