Skip to Content
Merck

52455

Sodium chloride physiological solution

BioUltra, tablet

Synonym(s):

Physiological Saline, tablets, Saline, physiological, tablets

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352302
EC Number:
231-598-3
MDL number:
Grade:
reagent grade
Form:
tablet
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

reagent grade

Quality Segment

product line

BioUltra

form

tablet

SMILES string

[Na]Cl

InChI

1S/ClH.Na/h1H;/q;+1/p-1

InChI key

FAPWRFPIFSIZLT-UHFFFAOYSA-M

Packaging

100 tablets in a bottle

Preparation Note

1 tablet in 1000 mL of water yields 0.9% NaCl


Still not finding the right product?


Storage Class

13 - Non Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library