Skip to Content

51912

Lewis-X trisaccharide 3′-sulfate sodium salt

≥95% (TLC)

Synonym(s):

Lex-sulfate sodium salt, SO3(O→3)-β-Gal-(1→4)(α-Fuc-[1→3])-GlcNAc, Sulfo-Lewis-X trisaccharide sodium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C20H34NO18SNa
CAS Number:
Molecular Weight:
631.53
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352201
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥95% (TLC)

form

solid

storage temp.

−20°C

SMILES string

[Na+].C[C@@H]1O[C@@H](O[C@@H]2[C@@H](NC(C)=O)C(O)O[C@H](CO)[C@H]2O[C@@H]3O[C@H](CO)[C@H](O)[C@H](OS([O-])(=O)=O)[C@H]3O)[C@@H](O)[C@H](O)[C@@H]1O

InChI

1S/C20H35NO18S.Na/c1-5-10(25)12(27)13(28)19(34-5)38-16-9(21-6(2)24)18(30)35-8(4-23)15(16)37-20-14(29)17(39-40(31,32)33)11(26)7(3-22)36-20;/h5,7-20,22-23,25-30H,3-4H2,1-2H3,(H,21,24)(H,31,32,33);/q;+1/p-1/t5-,7+,8+,9+,10+,11-,12+,13-,14+,15+,16+,17-,18?,19-,20-;/m0./s1

InChI key

QQNNDBNDALKSBB-LOBKVNCFSA-M



Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library