Skip to Content
Merck

51227

Dextran sulfate sodium salt

Mw 7,000-20,000

Synonym(s):

Dextran Sulfate Sodium

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.25
UNSPSC Code:
12352201
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

powder

Quality Segment

mol wt

Mw 7,000-20,000

loss

≤10% loss on drying

color

white

useful pH range

6.0-8 (10 g/L)

solubility

H2O: 100 mg/mL, clear, colorless to faintly greenish-yellow

storage temp.

room temp

General description

Dextran is a polymer of anhydroglucose. It is composed of approximately 95% alpha-D-(166) linkages. The remaining (163) linkages account for the branching of dextran.

Application

Dextran sulfate is routinely used for the selective precipitation of lipoproteins, probe hybridization membrane-immobilized DNA, the release of DNA from DNA-histone complexes, and the inhibition of RNA binding to ribosomes.

Biochem/physiol Actions

Dextran sulfate inhibits the replication of the human immunodeficiency virus by preventing the adsorption of the virus into host cells.

Other Notes

To gain a comprehensive understanding of our extensive range of Dextrans for your research, we encourage you to visit our Carbohydrates Category page.


Still not finding the right product?

Explore all of our products under Dextran sulfate sodium salt


Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library