Skip to Content
Merck

46947

Fluorescein isothiocyanate – Dextran 500000-Conjugate

(FITC:Glucose = 1:100)

Synonym(s):

Dextran 500′000 Fluorescein isothiocyanate Conjugate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.32
UNSPSC Code:
12352108
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

powder or crystals

Quality Segment

composition

(FITC:Glucose = 1:100)

extent of labeling

0.01 mol/mol FITC glucose (substitution)

technique(s)

permeability assay: suitable

solubility

H2O: 25 mg/mL, clear to very faintly turbid, yellow to orange

fluorescence

λex 490 nm; λem 520 nm (pH 8.0)

storage temp.

2-8°C

Application

Fluorescein isothiocyanate – Dextran 500000-Conjugate is used as a fluorescent tissue permeability assay reagent and as a diffusion probe for evaluation of biomaterials such as hydrogels and lipid vesicles.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library