Skip to Content
Merck

45252

Wright stain

BioReagent, suitable for microscopy

Synonym(s):

Eosin Methylene blue according to Wright

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
41116121
NACRES:
NA.25
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

BioReagent

Quality Segment

suitability

suitable for microscopy

SMILES string

S5c6c(ccc(c6)N(C)C)N=C7C5=CC(=[N+](C)C)C=C7.Brc1c2[o+]c3c(c(c2cc(c1O)Br)c4c(cccc4)C(=O)O)cc(c(c3Br)O)Br

InChI

1S/C20H8Br4O5.C16H18N3S/c21-11-5-9-13(7-3-1-2-4-8(7)20(27)28)10-6-12(22)17(26)15(24)19(10)29-18(9)14(23)16(11)25;1-18(2)11-5-7-13-15(9-11)20-16-10-12(19(3)4)6-8-14(16)17-13/h1-6H,(H2-,25,26,27,28);5-10H,1-4H3/q;+1/p+1

InChI key

AXIKDPDWFVPGOD-UHFFFAOYSA-O

Application

Reported to be superior to May-Grunwald Stain in blood staining.


Still not finding the right product?

Explore all of our products under Wright stain


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 3

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library