Skip to Content
Merck

44558

N-Octanoyl-L-homoserine lactone

≥96% (HPLC)

Synonym(s):

N-[(3S)-Tetrahydro-2-oxo-3-furanyl]octanamide, C8-HSL

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C12H21NO3
CAS Number:
Molecular Weight:
227.30
NACRES:
NA.85
PubChem Substance ID:
UNSPSC Code:
12352211
MDL number:
Beilstein/REAXYS Number:
8148282
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

N-Octanoyl-L-homoserine lactone, ≥96% (HPLC)

Quality Segment

assay

≥96% (HPLC)

form

powder or crystals

optical activity

[α]/D -28±3°, c = 0.2 in methanol

color

white to off-white

suitability

conforms to structure for Proton NMR spectrum

application(s)

detection

storage temp.

−20°C

SMILES string

O=C1OCC[C@@H]1NC(CCCCCCC)=O

InChI

1S/C12H21NO3/c1-2-3-4-5-6-7-11(14)13-10-8-9-16-12(10)15/h10H,2-9H2,1H3,(H,13,14)/t10-/m0/s1

InChI key

JKEJEOJPJVRHMQ-JTQLQIEISA-N

Biochem/physiol Actions

N-Octanoyl-L-homoserine lactone is a member of N-acyl-homoserine lactone family. N-Acylhomoserine lactones (AHL) regulate gene expression in gram-negative bacteria, such as Echerichia and Salmonella, and are involved in quorum sensing, colonization, chemical defense and cell to cell communication among bacteria; for reviews see. AHLs are released as oligopeptides in Gram positive bacteria and as quorum signal in Gram negative bacteria. Bacterial intercellular communication has become a target for the development of new anti-virulence drugs, and a research focus for the prevention of biofilm formation.
Quorum-sensing signal generation


Still not finding the right product?

Explore all of our products under N-Octanoyl-L-homoserine lactone


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
44558-50MG04061832283982
44558-10MG04061832283975