Skip to Content
Merck

Skip To

4445494

P450 Human Induction Starter Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.24
UNSPSC Code:
12352204
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


technique(s)

LC/MS: suitable

Quality Segment

General description

The P450 Human Induction Starter Kit provides a complete package for the analysis of four human CYP450 isoforms (1A2, 2B6, 3A4, and 3A5) using MRM analysis. The kit contains a detailed protocol, instrument data acquisition methods, processing methods, LC column, stable isotope-labeled synthetic peptide standards, trypsin, and all buffers and reagents for 100 assays.

Analysis Note

To view the Protocol for the CYP450 Protein Assay - Human Induction Kit, please visit this Protocol link.

To view the Quick Reference Card for the CYP450 Protein Assay - Human Induction Kit, please visit this Quick Reference Card link.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
44660044445252CLS4069
description

-

description

-

description

-

description

laboratory pipettor starter kit, w/ four Lambda Plus single-channel pipettors conveniently packaged w/ useful accessories, 1/cs

technique(s)

LC/MS: suitable

technique(s)

LC/MS: suitable

technique(s)

LC/MS: suitable

technique(s)

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

-


pictograms

Health hazard

signalword

Warning

hcodes

Hazard Classifications

STOT SE 2 Inhalation

Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters