Skip to Content
Merck
All Photos(1)

Key Documents

43781

Sigma-Aldrich

(Lauryldimethylammonio)acetate

≥95% (HPLC)

Synonym(s):

(Dodecyldimethylammonio)acetate, N-Dodecyl-N,N-dimethylglycine, Laurylbetaine, Lauryldimethylaminoacetic acid betaine, Lauryldimethylbetaine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H33NO2
CAS Number:
Molecular Weight:
271.44
Beilstein:
3670807
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95% (HPLC)

form

powder

mol wt

271.44 g/mol

technique(s)

HPLC: suitable

impurities

insoluble matter, passes filter test

conductivity

≤10 μS/cm (1% (w/v) in water)

λ

1 % (w/v) in H2O

UV absorption

λ: 260 nm Amax: ≤0.20
λ: 280 nm Amax: ≤0.15

SMILES string

CCCCCCCCCCCC[N+](C)(C)CC([O-])=O

InChI

1S/C16H33NO2/c1-4-5-6-7-8-9-10-11-12-13-14-17(2,3)15-16(18)19/h4-15H2,1-3H3

InChI key

DVEKCXOJTLDBFE-UHFFFAOYSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mariko Sugiyama et al.
The Journal of toxicological sciences, 44(6), 393-403 (2019-06-07)
To predict the results of a 24-hr closed human patch test, we previously recommended the use of in vitro test with a reconstructed human epidermis (RhE) model adopted in OECD TG 439, and proposed the margin method, which includes evaluation
M Machida et al.
Biological & pharmaceutical bulletin, 17(10), 1375-1378 (1994-10-01)
The effects of various surfactants and protease inhibitors on the nasal absorption of recombinant human granulocyte colony-stimulating factor (rhG-CSF) were examined in rats. No effects of bile salts and acids such as sodium glycocholate or taurocholic acid, amphoteric surfactants such
S Ishikawa et al.
Journal of applied microbiology, 93(2), 302-309 (2002-07-31)
In order to clarify the involvement of an energy-yielding system in the antibacterial action of surfactants, the effects of carbon source and anaerobiosis during the growth period on the surfactant sensitivity of Escherichia coli cells were investigated. Cetyltrimethylammonium bromide (CTAB)
C Combes et al.
Acta tropica, 39(1), 79-84 (1982-03-01)
Cercariae immobilisation and Schistosoma mansoni adult recovery tests in the mouse show that tensides can greatly reduce the infectivity of cercariae. In the case of two amphoteric products, Laurylbetain and Laurylamidopropylbetain, this action is obtained with very low (0.1-5 ppm)
Eric Vavra et al.
Scientific reports, 10(1), 12930-12930 (2020-08-02)
The apparent viscosity of viscous heavy oil emulsions in water can be less than that of the bulk oil. Microfluidic flooding experiments were conducted to evaluate how alkali-surfactant-foam enhanced oil recovery (ASF EOR) of heavy oil is affected by emulsion

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service