Skip to Content
Merck
All Photos(1)

Documents

43608

Sigma-Aldrich

3,3′-Dipropylthiadicarbocyanine iodide

suitable for fluorescence, ≥98.0% (TLC)

Synonym(s):

3-Propyl-2-[5-[3-propyl-2(3H)-benzothiazolylidene]-1,3-pentadienyl]benzothiazolium iodide, DiSC3(5)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H27IN2S2
CAS Number:
Molecular Weight:
546.53
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥98.0% (TLC)

form

powder

mp

245 °C (dec.) (lit.)

solubility

DMF: soluble

fluorescence

λex 500 nm; λem 705 nm in DMF

suitability

suitable for fluorescence

SMILES string

[I-].CCCN1C(\Sc2ccccc12)=C\C=C\C=C\c3sc4ccccc4[n+]3CCC

InChI

1S/C25H27N2S2.HI/c1-3-18-26-20-12-8-10-14-22(20)28-24(26)16-6-5-7-17-25-27(19-4-2)21-13-9-11-15-23(21)29-25;/h5-17H,3-4,18-19H2,1-2H3;1H/q+1;/p-1

InChI key

GDEURKKLNUGTDA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

3,3′-Dipropylthiadicarbocyanine iodide [DiSC3(5)] is used as a fluorogenic probe to detect and measure changes in transmembrane potential or configuration induced by membrane altering agents.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Transmembrane potential measurements

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wen Ma et al.
Journal of proteomics, 150, 242-251 (2016-10-25)
Daptomycin (DAP) is a novel lipopeptide antibiotic which exhibits excellent antibacterial activity against most clinically relevant Gram-positive bacteria, but the DAP-targeting protein molecules against host bacterial infection are far from clear. In order to discover bacterial protein response to DAP
M Síp et al.
Journal of photochemistry and photobiology. B, Biology, 4(3), 321-328 (1990-01-01)
The mechanism of carbocyanine dye diS-C3-(5) fluorescence intensity variations with transmembrane potential changes has been studied using time-resolved fluorescence spectroscopy. Clear evidence is given of the transmembrane-potential-dependent partition of the dye among various sites with different fluorescence lifetimes. It was
Min Kyoung Shin et al.
Antibiotics (Basel, Switzerland), 9(7) (2020-07-28)
The prevalence of antibiotic-resistant bacteria has become an immediate threat to public health. Antimicrobial peptides are attracting attention as a new source of antibiotics due to their ability to prevent drug-resistances with fewer side effects. Spider venom is composed of
Sonya E Neal et al.
Molecular biology of the cell, 28(21), 2773-2785 (2017-08-18)
Prokaryotes have aerobic and anaerobic electron acceptors for oxidative folding of periplasmic proteins. The mitochondrial intermembrane space has an analogous pathway with the oxidoreductase Mia40 and sulfhydryl oxidase Erv1, termed the mitochondrial intermembrane space assembly (MIA) pathway. The aerobic electron
Marie Markones et al.
Biophysical journal, 118(2), 294-302 (2019-12-18)
Membrane proteins are embedded in a complex lipid environment that influences their structure and function. One key feature of nearly all biological membranes is a distinct lipid asymmetry. However, the influence of membrane asymmetry on proteins is poorly understood, and

Articles

Nitric oxide (NO) as a signal transporter in neurons, endothelial cells and in the immune system.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service