42922
Micro particles based on polystyrene, dark red
size: 3 μm
Synonym(s):
Latex beads from PS, dark red
About This Item
Recommended Products
Quality Level
form
aqueous suspension
crosslinking
2 % cross-linked
concentration
5% (solids)
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service