Skip to Content
Merck

Skip To

39953

Lauroyl-L-carnitine

≥95.0% (HPLC)

Synonym(s):

(-)-Lauroylcarnitine, dodecanoylcarnitine, (2R)-3-Carboxy-N,N,N-trimethyl-2-[(1-oxododecyl)oxy]-1-propanaminium inner salt, L-Carnitine dodecanoyl ester, C12-Carnitine, Dodecanoyl-L-carnitine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C19H37NO4
CAS Number:
Molecular Weight:
343.50
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.26
Beilstein/REAXYS Number:
4147610
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95.0% (HPLC)

form

powder or crystals

optical activity

[α]/D -20±2°, c = 1 in methanol

packaging

pkg of 10 mg, pkg of 50 mg

impurities

≤10% water

color

white to off-white

functional group

ester

storage temp.

2-8°C

SMILES string

C[N+](C)(C)C[C@H](OC(CCCCCCCCCCC)=O)CC([O-])=O

InChI

1S/C19H37NO4/c1-5-6-7-8-9-10-11-12-13-14-19(23)24-17(15-18(21)22)16-20(2,3)4/h17H,5-16H2,1-4H3/t17-/m1/s1

InChI key

FUJLYHJROOYKRA-QGZVFWFLSA-N

General description

Lauroyl-L-carnitine, or Dodecanoylcarnitine, is an acylcarnitine compound with essential roles in energy metabolism. Synthesized in the liver from methionine, this metabolite of lysine is integral in various metabolic processes, making it a valuable asset in both biochemical and metabolomics research. Its concentration can serve as a diagnostic marker, particularly in cases of carnitine deficiency, and is of significance in assessing disease activity and diagnosing conditions like HIV and chronic hepatitis B infections. Additionally, Lauroyl-L-carnitine is associated with fatty acid oxidation disorders, including long-chain acyl CoA dehydrogenase deficiency and carnitine palmitoyltransferase I/II deficiency, further enhancing its relevance in metabolic research and diagnosis[1]. This acylcarnitine compound has also been detected in cancer tissues, human serum, and tissue biopsies, expanding its applications and potential contributions to various areas of metabolomics research.

Application

Lauroyl-L-carnitine finds application in biochemical research and metabolomics research.

Biochem/physiol Actions

Lauroyl-L-carnitine is an acylcarnitine is present in fatty acid oxidation disorders such as long-chain acyl CoA dehydrogenase deficiency, carnitine palmitoyltransferase I deficiency, and carnitine palmitoyltransferase II deficiency.

Features and Benefits

  • Can be used in Metabolomics and Biochemical research
  • High-quality compound suitable for multiple research applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
506375137151085
assay

≥95.0% (HPLC)

assay

≥94.0% (HPLC)

assay

≥94.0% (HPLC)

assay

≥97.0% (HPLC)

functional group

ester

functional group

ester

functional group

-

functional group

-

packaging

pkg of 10 mg, pkg of 50 mg

packaging

-

packaging

-

packaging

-

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

form

powder or crystals

form

powder or crystals

form

powder or crystals

form

powder

color

white to off-white

color

-

color

white to off-white

color

white


Still not finding the right product?

Explore all of our products under Lauroyl-L-carnitine


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters