Skip to Content
Merck

Skip To

36801

7-(Diethylamino)coumarin-3-carboxylic acid N-succinimidyl ester

BioReagent, suitable for fluorescence, ≥96.0% (HPLC)

Synonym(s):

N-Succinimidyl 7-(diethylamino)coumarin-3-carboxylate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C18H18N2O6
CAS Number:
Molecular Weight:
358.35
UNSPSC Code:
12352108
NACRES:
NA.32
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
5359433
Assay:
≥96.0% (HPLC)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

BioReagent

Quality Segment

assay

≥96.0% (HPLC)

solubility

DMF: soluble, DMSO: soluble, acetonitrile: soluble

fluorescence

λex 445 nm; λem 482 nm in 0.1 M phosphate pH 7.0

suitability

corresponds for coupling to amines, suitable for fluorescence

storage temp.

−20°C

SMILES string

CCN(CC)c1ccc2C=C(C(=O)ON3C(=O)CCC3=O)C(=O)Oc2c1

InChI

1S/C18H18N2O6/c1-3-19(4-2)12-6-5-11-9-13(17(23)25-14(11)10-12)18(24)26-20-15(21)7-8-16(20)22/h5-6,9-10H,3-4,7-8H2,1-2H3

InChI key

NUNPVRICKDZFLK-UHFFFAOYSA-N

Application

7-(Diethylamino)coumarin-3-carboxylic acid N-succinimidyl ester is used as a reagent to conjugate 7-(diethylamino)coumarin-3-carboxylic acid (7-DCCA) to other molecules such as amino acids via amide chemistry. 7-DCCA is used to label amino acids prior to resolution by capillary zone electrophoresis. 7-DCCA is used as a reference in screening of nerve agent degradation products by MALDI-TOFMS.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Labeling of amino acids prior to resolution by capillary zone electrophoresis

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
55156M979487444
description

BioReagent, suitable for fluorescence, ≥96.0% (HPLC)

description

suitable for fluorescence, ≥95.0% (coupling assay to aminopropyl silica gel)

description

≥98%, powder

description

suitable for fluorescence, ≥95.0% (HPLC)

assay

≥96.0% (HPLC)

assay

≥95.0% (coupling assay to aminopropyl silica gel)

assay

≥98%

assay

≥95.0% (HPLC)

solubility

DMF: soluble, acetonitrile: soluble, DMSO: soluble

solubility

DMF: soluble, DMSO: soluble

solubility

chloroform: 100 mg/mL, DMF: soluble (lit.)(lit.)

solubility

DMF: soluble, DMSO: soluble

Quality Level

200

Quality Level

100

Quality Level

200

Quality Level

100

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

fluorescence

λex 445 nm; λem 482 nm in 0.1 M phosphate pH 7.0

fluorescence

λex 386 nm; λem 448 nm in 0.1 M Tris pH 9.0

fluorescence

-

fluorescence

λex 492 nm; λem 517 nm

suitability

corresponds for coupling to amines, suitable for fluorescence

suitability

-

suitability

-

suitability

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters