Skip to Content
Merck

317616

D.P.X.

Neutral mounting medium

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12171500
EC Number:
215-535-7
NACRES:
NA.47
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic

Quality Segment

form

viscous liquid

color

yellow

refractive index

n20/D 1.521 (lit.)

density

0.904 g/mL at 25 °C (lit.)

application(s)

hematology
histology

storage temp.

room temp

General description

D.P.X is a mixture of distyrene (polystyrene), a plasticizer (tricresyl phosphate), and xylene. It facilitates the protection of stains and can dry quickly.

Application

D.P.X. has been used as a mounting medium for mounting:
  • rat brain sections for immunohistochemistry studies
  • mouse small intestine sections for morphometry studies
  • gastrointestinal tract sections of rats for immunohistochemistry



signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

target_organs

Central nervous system,Liver,Kidney, Respiratory system

Storage Class

3 - Flammable liquids

wgk

WGK 3

flash_point_f

77.0 °F - closed cup

flash_point_c

25 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library