Skip to Content
Merck
All Photos(1)

Key Documents

30800

Sigma-Aldrich

7-Dehydrocholesterol

≥95.0% (HPLC)

Synonym(s):

(−)-7-Dehydrocholesterol, 3β-Hydroxy-5,7-cholestadiene, 5,7-Cholestadien-3β-ol, Provitamin D3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H44O
CAS Number:
Molecular Weight:
384.64
Beilstein:
2224615
EC Number:
MDL number:
UNSPSC Code:
12352211
eCl@ss:
39023139
PubChem Substance ID:
NACRES:
NA.79

biological source

synthetic

Assay

≥95.0% (HPLC)

form

powder or crystals

optical activity

[α]20/D -115±8°, c = 1% in chloroform

color

white to faint yellow

mp

148-152 °C (lit.)

storage temp.

−20°C

SMILES string

[H][C@@]1(CC[C@@]2([H])C3=CC=C4C[C@@H](O)CC[C@]4(C)[C@@]3([H])CC[C@]12C)[C@H](C)CCCC(C)C

InChI

1S/C27H44O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h9-10,18-19,21,23-25,28H,6-8,11-17H2,1-5H3/t19-,21+,23-,24+,25+,26+,27-/m1/s1

InChI key

UCTLRSWJYQTBFZ-DDPQNLDTSA-N

Looking for similar products? Visit Product Comparison Guide

General description

7-Dehydrocholesterol (7-DHC), a 5,7-conjugated diene sterol is a biosynthetic precursor of cholesterol. It helps in the production of vitamin D3 when exposed to ultraviolet B (UVB) radiation.

Application

7-Dehydrocholesterol has been used as an internal standard to determine sterols.

Biochem/physiol Actions

Down-regulates cholesterol biosynthesis in cultured Smith-Lemi-Opitz syndrome skin fibroblasts.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amaranth oil increased fecal excretion of bile acid but had no effect in reducing plasma cholesterol in hamsters
de Castro lIA, et al.
Lipids, 48, 609-618 (2013)
7-Dehydrocholesterol (7-DHC), But Not Cholesterol, Causes Suppression of Canonical TGF-beta Signaling and Is Likely Involved in the Development of Atherosclerotic Cardiovascular Disease (ASCVD)
Huang SS, et al.
Journal of Cellular Biochemistry, 118(6), 1387-1387 (2017)
P E Jira et al.
Journal of lipid research, 41(8), 1339-1346 (2000-08-18)
The Smith-Lemli-Opitz syndrome (SLOS) is caused by deficient Delta(7)-dehydrocholesterol reductase, which catalyzes the final step of the cholesterol biosynthetic pathway, resulting in low cholesterol and high concentrations of its direct precursors 7-dehydrocholesterol (7DHC) and 8DHC. We hypothesized that i) 7DHC
Daniela Criscuolo et al.
Cells, 9(4) (2020-04-03)
Despite initial chemotherapy response, ovarian cancer is the deadliest gynecologic cancer, due to frequent relapse and onset of drug resistance. To date, there is no affordable diagnostic/prognostic biomarker for early detection of the disease. However, it has been recently shown
Vitamin D, Calcium, and the Epidermis
Vitamin D, 1, 527-544 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service