Skip to Content
Merck
All Photos(1)

Key Documents

27606

Sigma-Aldrich

DL-Malic acid

meets analytical specification of FCC, E296, 99-100.5% (alkalimetric)

Synonym(s):

(±)-2-Hydroxysuccinic acid, DL-Hydroxybutanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCH2CH(OH)CO2H
CAS Number:
Molecular Weight:
134.09
Beilstein:
1723539
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

vapor density

4.6 (vs air)

Quality Level

vapor pressure

<0.1 mmHg ( 20 °C)

Assay

99-100.5% (alkalimetric)

form

solid

optical activity

[α]/D −0.10 to +0.10°

autoignition temp.

644 °F

quality

meets analytical specification of FCC, E296

technique(s)

IR spectroscopy: suitable

impurities

≤0.05% maleic acid
≤0.1% water insoluble matter
≤1% fumaric acid

ign. residue

≤0.1% (as SO4)

mp

129-133 °C
131-133 °C (lit.)

solubility

acetone: soluble 17.75 g/ 100 gm at 20 °C
diethyl ether: soluble 0.84 g/100 gm at 20 °C
dioxane: soluble 22.70 g/100 gm at 20 °C
ethanol: soluble 45.53 g/100 gm at 20 °C
methanol: soluble 82.70 g/100 g at 20 °C
water: soluble 55.8 g/100 gm at 20 °C
benzene: insoluble

cation traces

As: ≤3 mg/kg
Hg: ≤1 mg/kg
Pb: ≤2 mg/kg

suitability

complies for identity (IR)

application(s)

pharmaceutical (small molecule)

SMILES string

OC(CC(O)=O)C(O)=O

InChI

1S/C4H6O5/c5-2(4(8)9)1-3(6)7/h2,5H,1H2,(H,6,7)(H,8,9)

InChI key

BJEPYKJPYRNKOW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

DL-Malic acid is an organic acid used for studying the impact on ultrasound pasteurization.[1] It has also been used to culture Pichia membraneafaciens.[2]

Biochem/physiol Actions

Malic acid is a dicarboxylic acid and an important regulatory metabolite. It has been implicated in process of fruit ripening. Malic acid is important for the starch metabolism; low malic acid content results in transient accumulation of starch. Mitochondrial-malate metabolism modulates ADP-glucose pyrophosphorylase activity and redox status of plastids.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

397.4 °F

Flash Point(C)

203 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L-Malic acid production by polyacrylamide gel entrapped Pichia membranaefaciens.
Biotechnology Letters, 7, 329-334 (1985)
S Z Salleh-Mack et al.
Ultrasonics sonochemistry, 14(3), 323-329 (2006-09-13)
The objectives of this research were to study the effects of temperature, sugar concentration (8, 12, and 16 g/100ml), organic acids (citric and malic acids) and pH (2.5 and 4.0) on ultrasound pasteurization. The model organism used for the research
Danilo C Centeno et al.
The Plant cell, 23(1), 162-184 (2011-01-18)
Despite the fact that the organic acid content of a fruit is regarded as one of its most commercially important quality traits when assessed by the consumer, relatively little is known concerning the physiological importance of organic acid metabolism for
G Anneroth et al.
Scandinavian journal of dental research, 88(3), 214-218 (1980-06-01)
Two saliva stimulating drugs, solutions of Hybrin and malic acid were in an in vitro study found to have a heavy demineralizing effect on the root surface of the dental hard tissue. Consequently the drugs cannot be recommended to patients
Hugo Oliveira et al.
PloS one, 9(10), e108376-e108376 (2014-10-08)
Resistance rates are increasing among several problematic Gram-negative pathogens, a fact that has encouraged the development of new antimicrobial agents. This paper characterizes a Salmonella phage endolysin (Lys68) and demonstrates its potential antimicrobial effectiveness when combined with organic acids towards

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service