24531C
EX-CELL® EBx™ PRO-I Serum-Free Medium for Embryonic Stem Cells
without L-Glutamine, without Phenol Red, without Sodium Bicarbonate
Pharma Manufacturing
Synonym(s):
EX-CELL® EBx™ Serum-Free Media
About This Item
description
Animal-component-free, low protein
for research or for further manufacturing use
Quality Level
form
dry powder
technique(s)
cell culture | mammalian: suitable
storage temp.
2-8°C
2-8°C
Looking for similar products? Visit Product Comparison Guide
Quantity
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Sens. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service