Skip to Content
Merck
All Photos(1)

Documents

244R-1

Sigma-Aldrich

DOG1 (SP31) Rabbit Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

SP31, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (244R-14)
vial of 0.5 mL concentrate (244R-15)
bottle of 1.0 mL predilute (244R-17)
vial of 1.0 mL concentrate (244R-16)
bottle of 7.0 mL predilute (244R-18)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

isotype

IgG

control

GIST

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic, membranous

Gene Information

human ... ANO1(55107)

General description

DOG1 is a calcium-dependent chloride channel protein that is encoded by a gene called TMEM16A (TMEM16 FLJ10261, ANO1, ORAOV2, and AOS2) located on chromosome 11q13. DOG1 has many significant functions such as regulation of the cholinergic activity of gastrointestinal smooth muscle and regulation of both the survival and proliferation of cells. Anti-DOG1 antibody has been shown to be useful in the identification of gastrointestinal stromal tumors (GIST).

Quality


IVD

IVD

IVD

RUO

Linkage

DOG1 Positive Control Slides, Product No. 244S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J R Parfitt et al.
Histopathology, 52(7), 816-823 (2008-05-23)
Gastrointestinal Kaposi's sarcoma (KS) may mimic gastrointestinal stromal tumours (GISTs) histologically. Studies have shown that KS outside the gastrointestinal (GI) tract may express CD117, an antibody usually used to support a diagnosis of GIST. The aim was to evaluate the
Robert B West et al.
The American journal of pathology, 165(1), 107-113 (2004-06-25)
We recently characterized gene expression patterns in gastrointestinal stromal tumors (GISTs) using cDNA microarrays, and found that the gene FLJ10261 (DOG1, discovered on GIST-1), encoding a hypothetical protein, was specifically expressed in GISTs. The immunoreactivity of a rabbit antiserum to
Shigeharu Miwa et al.
Journal of gastroenterology, 43(7), 531-537 (2008-07-24)
Recently, the novel gene DOG1 has been found to be overexpressed in most gastrointestinal stromal tumors (GISTs) specifically within the field of soft tissue tumors. DOG1 might play a role in development of GISTs and have potential as a diagnostic
Inigo Espinosa et al.
The American journal of surgical pathology, 32(2), 210-218 (2008-01-29)
Gastrointestinal stromal tumors (GIST) occur primarily in the wall of the intestine and are characterized by activating mutations in the receptor tyrosine kinases genes KIT or PDGFRA. The diagnosis of GIST relies heavily on the demonstration of KIT/CD117 protein expression

Articles

Colorectal cancer is the third most common cancer in both men and women. An estimated 136,000 cases of colorectal cancer are expected to occur in 2016.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service