Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C29H25N3O7
CAS Number:
Molecular Weight:
527.52
UNSPSC Code:
12352116
NACRES:
NA.32
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
8739319
product line
BioReagent
Quality Level
assay
≥70% (coupling to amines)
form
powder
solubility
DMF: soluble, acetonitrile: soluble, methanol: soluble
fluorescence
λex 543 nm; λem 576 nm in methanol, λex 554 nm; λem 584 nm in 0.1 M phosphate pH 8.0
suitability
suitable for fluorescence
storage temp.
−20°C
SMILES string
CN(C)c1ccc2c(Oc3cc(ccc3C24OC(=O)c5cc(ccc45)C(=O)ON6C(=O)CCC6=O)N(C)C)c1
InChI
1S/C29H25N3O7/c1-30(2)17-6-9-21-23(14-17)37-24-15-18(31(3)4)7-10-22(24)29(21)20-8-5-16(13-19(20)28(36)38-29)27(35)39-32-25(33)11-12-26(32)34/h5-10,13-15H,11-12H2,1-4H3
InChI key
CXYYHBMOVJJZTD-UHFFFAOYSA-N
Application
5(6)-Carboxytetramethylrhodamine N-succinimidyl ester (TMR-SE) is used as an amine coupling reagent to form 5(6)-carboxytetramethylrhodamine (TMR) derivatized compounds such as proteins and drugs.
Packaging
Bottomless glass bottle. Contents are inside inserted fused cone.
Other Notes
Rhodamine derived label for amine modification and protein conjugation
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
L D Mayfield et al.
Bioorganic & medicinal chemistry letters, 9(10), 1419-1422 (1999-06-09)
Resins for solid-phase synthesis can affect coupling efficiencies by interacting with reactants. We have observed that polyethylene glycol-polystyrene (PEG-PS) solid support absorbs added activated fluorophores, preventing efficient labeling of peptide nucleic acids (PNAs). We now report that addition of an
E. Koller
Appl. Fluoresc. Technol., 3, 20-20 (1991)
Divyamani Srinivasan et al.
PloS one, 6(3), e17732-e17732 (2011-03-23)
Cell-penetrating peptides (CPPs) can transport macromolecular cargos into live cells. However, the cellular delivery efficiency of these reagents is often suboptimal because CPP-cargo conjugates typically remain trapped inside endosomes. Interestingly, irradiation of fluorescently labeled CPPs with light increases the release
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 21955-25MG | 04061838776020 |