Skip to Content
Merck
All Photos(1)

Key Documents

21605

Sigma-Aldrich

Patent blue V sodium salt

for microscopy

Synonym(s):

Acid blue 3 sodium salt, Food Blue 5 sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H31N2NaO7S2
CAS Number:
Molecular Weight:
582.66
Colour Index Number:
42051
Beilstein:
4108121
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

grade

for microscopy

Assay

5 — 10% Na basis

form

powder

technique(s)

microbe id | staining: suitable

color

blue to very dark blue

pH range

4.0-6.0

solubility

methanol: water (1:1): 0.01 g/10 mL, blue to very deep blue

εmax

≥1200 at 630-640 nm in water
≥50 at 405-415 nm in water

suitability

suitable for microscopy

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].CCN(CC)c1ccc(cc1)\C(=C2/C=CC(\C=C2)=[N+](\CC)CC)c3cc(O)c(cc3S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/C27H32N2O7S2.Na/c1-5-28(6-2)21-13-9-19(10-14-21)27(20-11-15-22(16-12-20)29(7-3)8-4)23-17-24(30)26(38(34,35)36)18-25(23)37(31,32)33;/h9-18H,5-8H2,1-4H3,(H2-,30,31,32,33,34,35,36);/q;+1/p-1

InChI key

PMLFOMWMYRKZRF-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Patent Blue V sodium salt, also called as Sulfan Blue. is a disulfonated diaminotriphenylmethane dye with a hydrophilic anion and an aromatic system. It is orange under acid conditions and blue in alkali.

Application

Patent Blue V sodium salt has been used as a stain in pharmacology.It is a biological stain used for the demonstration of hemoglobin. Its applications are also seen in the field of biology and medicine where it′s used as a physiological tracer, to study the movement of fluid within the kidney. Patent Blue V is useful as a marker dye in oncological surgeries for accurate and complete excision of lymph nodes. It is also used as a food color.
Patent blue V sodium salt has been used as a stain in pharmacology.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Encyclopedia of Food and Color Additives, 3 (1997)
Tammo Krause et al.
Current biology : CB, 29(11), 1833-1841 (2019-05-21)
Grasping an object or crossing a trench requires the integration of information on the operating distance of our limbs with precise distance estimation. The reach of our hands and step size of our legs are learned by the visual feedback
Serotonin modulates a depression-like state in Drosophila responsive to lithium treatment
Ries AS
Nature Communications (2017)
Ting Chen et al.
iScience, 23(9), 101533-101533 (2020-10-22)
Lanthanide-based, Förster resonance energy transfer (LRET) biosensors enabled sensitive, time-gated luminescence (TGL) imaging or multiwell plate analysis of protein-protein interactions (PPIs) in living cells. We prepared stable cell lines that expressed polypeptides composed of an alpha helical linker flanked by
Agnes Rodler et al.
Journal of biotechnology, 288, 48-54 (2018-10-14)
Alternative separation methods operating in an aqueous environment are of increasing importance for further progress of molecular separation in life sciences and other industrial sectors working towards a biobased economy. By spincoating, membranes with thicknesses under 100 nm and 20

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service