Skip to Content
Merck

21054

Alginic acid calcium salt from brown algae

Synonym(s):

Calcium alginate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352201
NACRES:
NA.25
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

algae (brown)

Quality Segment

form

powder

ign. residue

~33% (as SO4)

loss

~10% loss on drying

color

light gray

application(s)

advanced drug delivery

storage temp.

room temp

SMILES string

[Ca+2].[O-]C(=O)[C@@H]1O[C@H]([C@H]([C@H]([C@@H]1O[C@@H]2O[C@H]([C@H]([C@@H]([C@@H]2O)O)O[C@@H]3O[C@@H]([C@H]([C@@H]([C@@H]3O)O)O)C(=O)[O-])C(=O)O)O)O)O

InChI

1S/C18H26O19.Ca/c19-1-2(20)10(13(26)27)36-17(6(1)24)35-9-4(22)7(25)18(37-12(9)15(30)31)34-8-3(21)5(23)16(32)33-11(8)14(28)29;/h1-12,16-25,32H,(H,26,27)(H,28,29)(H,30,31);/q;+2/p-2/t1-,2-,3+,4+,5-,6-,7-,8-,9-,10-,11+,12+,16+,17+,18+;/m0./s1

InChI key

OKHHGHGGPDJQHR-YMOPUZKJSA-L

Application

Alginate, a colloidal polyuronic acid structural molecule capable of gelation, is used in the preparation of colloidal biodegradable structures such as gels, biofilms, beads, nanoparticles, and microcapsules suitable for applications that range from gel based separation technologies to drug delivery and cell preservation.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library