Skip to Content
Merck

Skip To

202M-9

Actin, Smooth Muscle (1A4) Mouse Monoclonal Antibody

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
1A4, monoclonal
Application:
IHC (p)
Citations:
7
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

1A4, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (202M-94), vial of 0.5 mL concentrate (202M-95), bottle of 1.0 mL predilute (202M-97), vial of 1.0 mL concentrate (202M-96), bottle of 7.0 mL predilute (202M-98)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

isotype

IgGκ

control

appendix

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

General description

Smooth Muscle Actin is a part of the actin family of proteins which are highly conserved andform microfilaments. These filaments are one of the major components of the cytoskeleton.Anti-smooth muscle actin immunohistochemical reactivity is seen in smooth muscle cells,myofibroblasts, and myoepithelial cells.

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Analysis Note


IVD

IVD

IVD

RUO

Other Notes

Actin, Smooth Muscle Positive Control Slides, Product No. 202S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).
For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
CBL171-IABT1487A5228
species reactivity

human

species reactivity

mouse, chicken, rat, human, bovine

species reactivity

rat, mouse, human

species reactivity

human, mouse, rat, chicken, frog, canine, rabbit, guinea pig, goat, bovine, sheep, snake

biological source

mouse

biological source

mouse

biological source

rabbit

biological source

mouse

conjugate

unconjugated

conjugate

-

conjugate

-

conjugate

unconjugated

clone

1A4, monoclonal

clone

ASM-1 (1A4), monoclonal

clone

polyclonal

clone

1A4, monoclonal

antibody form

culture supernatant

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

purified immunoglobulin

form

buffered aqueous solution

form

-

form

-

form

PBS solution


Still not finding the right product?

Try our Product Selector Tool to narrow your options




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. Can Smooth Muscle Actin (1A4) be run on the Ventana/Roche Ultra using OptiView detection, and what would be the protocol for this?

    1 answer
    1. The protocol for this detection to run this antibody in that manner. The protocol available is with UltraView detection and No Pretreatment, a 32-minute antibody incubation at room temperature, plus amplifier.

      Helpful?

Reviews

No rating value

Active Filters