Skip to Content

1804

Pararosaniline Solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.47
UNSPSC Code:
41116124
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solution

Quality Segment

shelf life

Expiry date on the label.

IVD

for in vitro diagnostic use

concentration

40 g/L

application(s)

hematology
histology

storage temp.

room temp

Application

When mixed with a 4% sodium nitrite solution, a freshly hexazotized pararosaniline solution is produced. May be used in a variety of cytochemistry procedures, including Sigma Procedures 181B and 181C.

Other Notes

Pararosaniline, 40g/L, in 2 mol/L hydrochloric acid.


signalword

Danger

Hazard Classifications

Carc. 1B - Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library