Skip to Content
Merck
All Photos(1)

Key Documents

17936

Sigma-Aldrich

1α,25-Dihydroxyvitamin D3

≥97.0% (HPLC)

Synonym(s):

1α,25-Dihydroxycholecalciferol, Calcitriol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H44O3
CAS Number:
Molecular Weight:
416.64
Beilstein:
7559394
EC Number:
MDL number:
UNSPSC Code:
12352205
eCl@ss:
34058003
PubChem Substance ID:
NACRES:
NA.79

Quality Level

Assay

≥97.0% (HPLC)

form

crystals

color

white to off-white

storage temp.

−70°C

SMILES string

[H][C@@]1(CC[C@@]2([H])C(\CCC[C@]12C)=C\C=C3\C[C@@H](O)C[C@H](O)C3=C)[C@H](C)CCCC(C)(C)O

InChI

1S/C27H44O3/c1-18(8-6-14-26(3,4)30)23-12-13-24-20(9-7-15-27(23,24)5)10-11-21-16-22(28)17-25(29)19(21)2/h10-11,18,22-25,28-30H,2,6-9,12-17H2,1,3-5H3/b20-10+,21-11-/t18-,22-,23-,24+,25+,27-/m1/s1

InChI key

GMRQFYUYWCNGIN-NKMMMXOESA-N

Gene Information

human ... VDR(7421)

Looking for similar products? Visit Product Comparison Guide

General description

1α,25-Dihydroxyvitamin D3, also referred as calcitriol, is a calcitrophic hormone. It is the most biologically active metabolite of vitamin D produced in proximal tubular cells in the kidney.

Application

1α,25-Dihydroxyvitamin D3 has been used to study its effect on transplantation success of human muscle precursor cells (hMPCs) in severe combined immunodeficient (SCID) mice in-vitro.

Biochem/physiol Actions

1α,25-Dihydroxyvitamin D3 modulates energy metabolism by facilitating various metabolic processes, such as Ca2+ signaling and modulation of glucocorticoids, by affecting adipocyte apoptosis and by regulating the expression and production of multiple cytokines. In addition, it also modulates phosphorus homeostasis through actions in the intestine, kidney and bone. 1α,25-Dihydroxyvitamin D3 exhibits antiproliferative effects and is used as a potent therapeutic for human prostate cancer.
Biologically active form of vitamin D3 in calcium absorption and deposition.
Biologically active form of vitamin D3 in calcium absorption and deposition. 1α,25-Dihydroxyvitamin D3 has widespread effects on cellular differentiation and proliferation, and can modulate immune responsiveness, and central nervous system function. Recent studies suggest that 1α,25-dihydroxyvitamin D3 acts as a chemopreventive agent against several malignancies including cancers of the prostate and colon and shows synergy with other anticancer compounds.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Oral - Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - STOT RE 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Handbook of Hormones: Comparative Endocrinology for Basic and Clinical Research (2015)
Physiology of the Gastrointestinal Tract (2012)
Calcitriol and energy metabolism
Zemel M B and Sun X
Nutr. Rev, 66(suppl-2), S139-S146 (2008)
Treatment of early recurrent prostate cancer with 1, 25-dihydroxyvitamin D3 (calcitriol)
Gross C, et al.
The Journal of Urology, 159(6), 2035-2040 (1998)
Inhibition of T lymphocyte mitogenesis by 1, 25-dihydroxyvitamin D3 (calcitriol).
Rigby WF, et al.
The Journal of Clinical Investigation, 74(4), 1451-1455 (1984)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service