Skip to Content
Merck
All Photos(1)

Key Documents

17304

Sigma-Aldrich

Crocin

for microscopy

Synonym(s):

Crocetin digentiobiose ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H64O24
CAS Number:
Molecular Weight:
976.96
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.47

grade

for microscopy

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

SMILES string

CC(=C\C=C\C=C(C)\C=C\C=C(/C)C(=O)O[C@@H]1O[C@H](CO[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@H](O)[C@H]1O)\C=C\C=C(\C)C(=C)CO[C@@H]3O[C@H](CO[C@@H]4O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4O)[C@@H](O)[C@H](O)[C@H]3O

InChI

1S/C46H68O23/c1-21(12-8-14-23(3)25(5)18-62-43-40(59)36(55)32(51)28(67-43)19-63-44-38(57)34(53)30(49)26(16-47)65-44)10-6-7-11-22(2)13-9-15-24(4)42(61)69-46-41(60)37(56)33(52)29(68-46)20-64-45-39(58)35(54)31(50)27(17-48)66-45/h6-15,26-41,43-60H,5,16-20H2,1-4H3/b7-6+,12-8+,13-9+,21-10-,22-11+,23-14-,24-15+/t26-,27-,28-,29-,30-,31-,32-,33-,34+,35+,36+,37+,38-,39-,40-,41-,43-,44-,45-,46+/m1/s1

InChI key

LUVDBMJRTUBHNX-RSXXRLSLSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Crocin (CR) is a water-soluble carotenoid found in Gardenia jasmi-noides Ellis and Crocus sativus (saffron). It has two D-gentiobioside glycosyl esters in its structure. CR is one of the major bioactive compounds of saffron contributing to its red color.
A highly unsaturared diterpene dicarboxylic acid ester found in saffron.

Application

Crocin has been used:
  • as a reference standard to detect its presence in Crocus sativus L. extract using high-performance liquid chromatography technique
  • to investigate its protective effects as an antioxidant and anti-inflammatory agent against doxorubicin-induced nephrotoxicity in rats
  • to test antitumor effects and its role in promoting autophagy and apoptosis and inhibiting the progression of cervical cancer on SiHa cells and female BALB/c nude mice
  • as a co-treatment with cisplatin to test its protective effect against cisplatin (CIS)-induced testicular toxicity in rats

Biochem/physiol Actions

Crocin alleviates some ethanol-induced impairments of learning and prevents ethanol-induced inhibition of hippocampal long-term potentiation (LTP), a form of activity-dependent synaptic plasticity that may underly learning and memory. Related CNS effects are specific to the digentiobiose ester; crocetin glucose gentiobiose ester is half as potent, and the diglucose ester has no effect at all.
Crocin exerts antioxidant, anti-inflammation, anti-cancer, antidegenerative, and antidiabetic properties.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Fogelholm et al.
Journal of neurology, neurosurgery, and psychiatry, 55(12), 1121-1124 (1992-12-01)
Forty one (14.2%) of 288 patients with primary intracerebral haemorrhage occurring between September 1985 and December 1989 in Central Finland were on anticoagulant treatment at the onset of symptoms. In a sample of 29,000 subjects from the same population the
Olga Bountagkidou et al.
Journal of chromatography. A, 1237, 80-85 (2012-03-31)
An on-line HPLC (high performance liquid chromatography) method for the rapid screening of individual antioxidants in mixtures was developed using crocin as a substrate (i.e. oxidation probe) and 2,2'-azobis(2-amidinopropane dihydrochloride (AAPH)) in phosphate buffer (pH 7.5) as a radical generator.
Mika Masaki et al.
Molecular nutrition & food research, 56(2), 304-308 (2011-11-01)
Crocus sativus L. (saffron) has been traditionally used for the treatment of insomnia and other diseases of the nervous systems. Two carotenoid pigments, crocin and crocetin, are the major components responsible for the various pharmacological activities of C. sativus L.
Francesca Gullo et al.
Frontiers in neuroscience, 11, 500-500 (2017-09-22)
Increasing evidence supports a decisive role for neuroinflammation in the neurodegenerative process of several central nervous system (CNS) disorders. Microglia are essential mediators of neuroinflammation and can regulate a broad spectrum of cellular responses by releasing reactive oxygen intermediates, nitric
Amir Faramarzpour et al.
Veterinary research forum : an international quarterly journal, 10(3), 227-234 (2019-11-19)
Crocin, as a carotenoid compound of saffron, exerts a potent antioxidant property. Mesalazine is frequently used in the treatment of ulcerative colitis. This study investigated the effects of separated and combination treatments with crocin and mesalazine in a rat model

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service