Skip to Content
Merck
All Photos(1)

Key Documents

168M-9

Sigma-Aldrich

CD68 (Kp-1) Mouse Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

Kp-1, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (168M-94)
vial of 0.5 mL concentrate (168M-95)
bottle of 1.0 mL predilute (168M-97)
vial of 1.0 mL concentrate (168M-96)
bottle of 7.0 mL predilute (168M-98)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

isotype

IgG1κ

control

tonsil

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

Gene Information

human ... CD68(968)

General description

Anti-CD68 marks cells of monocyte/macrophage lineage. This antibody is capable of staining monocytes, Kupffer cells, osteoclasts, granulocytes and their precursors; lymphomas are negative or show a few granules. This antibody may be useful for the identification of myelomonocytic and histiocytic tumors. Anti-CD68 may help to distinguish malignant fibrous histiocytoma from other pleomorphic sarcomas. However, since this detects a formalin-resistant epitope that may be associated with lysosomal granules, other lysosome-rich cells may also stain.

Quality


IVD

IVD

IVD

RUO

Linkage

CD68 Positive Control Slides, Product No. 168S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J L Snow et al.
The British journal of dermatology, 133(1), 71-76 (1995-07-01)
The clinical, histopathological, and immunophenotypic characteristics of four cases of malignancy-associated multicentric reticulohistiocytosis (MMR) and one case each of diffuse cutaneous reticulohistiocytosis (DCR) and isolated reticulohistiocytoma (IR), are reviewed. In all four cases of MMR the cutaneous lesions and joint
B Vergier et al.
Blood, 95(7), 2212-2218 (2000-03-25)
The course of mycosis fungoides (MF) is indolent except when transformation to a large T-cell lymphoma occurs. The diagnosis of transformed MF (T-MF) relies on the presence of more than 25% of large cells on biopsy of an MF lesion.
L Martinez-Pomares et al.
Immunobiology, 195(4-5), 407-416 (1996-10-01)
Monoclonal antibodies directed against murine macrophage differentiation antigens provide tools to characterize novel glycoproteins, their expression by different macrophage subpopulations in situ and regulation by cytokines and other agents in vitro. Studies are in progress to determine ligands for these
CD68 cluster workshop report
JL Cordell, et al.
Leucocyte Typing V, 925-927 (1995)
L P Ruco et al.
American journal of clinical pathology, 92(3), 273-279 (1989-09-01)
The expression of macrophage antigens KP1, Mac, lysozyme, and alpha-1-antichymotrypsin was investigated on routine paraffin sections from 17 cases of Langerhans' cell histiocytosis (LCH). All the major clinical forms were represented, including single lesions and monosystemic and multisystemic disease. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service