Skip to Content

15090

1,4-Bis(2-methylstyryl)benzene

BioReagent, suitable for scintillation, ≥98% (UV)

Synonym(s):

Bis-MSB

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(CH3C6H4CH=CH)2C6H4
CAS Number:
Molecular Weight:
310.43
UNSPSC Code:
12352103
NACRES:
NA.32
PubChem Substance ID:
EC Number:
236-285-5
Beilstein/REAXYS Number:
2375845
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

BioReagent

Quality Segment

assay

≥98% (UV)

form

solid

mol wt

310.43 g/mol

composition

Carbon content (theory), 92.86% , Hydrogen content (theory), 7.14%

color

faint green to green, white to yellow

mp

180-182 °C

solubility

dioxane: 0.5 g/10 mL at hot, clear to hazy, colorless to light yellow

fluorescence

λem 416 nm in heptane, λem 419 nm in cyclohexane

suitability

suitable for scintillation

SMILES string

Cc1ccccc1\C=C\c2ccc(cc2)\C=C\c3ccccc3C

InChI

1S/C24H22/c1-19-7-3-5-9-23(19)17-15-21-11-13-22(14-12-21)16-18-24-10-6-4-8-20(24)2/h3-18H,1-2H3/b17-15+,18-16+

InChI key

QKLPIYTUUFFRLV-YTEMWHBBSA-N

Application

Absolute fluorescence emission in cyclohexane (max): 420nm. Utilized as a scintillator reagent.


Still not finding the right product?

Explore all of our products under 1,4-Bis(2-methylstyryl)benzene


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4 - Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library