Skip to Content
Merck

15029

Caftaric acid

≥97.0%

Synonym(s):

2-Caffeoyl-L-tartaric acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C13H12O9
CAS Number:
Molecular Weight:
312.23
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352106
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥97.0%

form

powder

impurities

≤10% water

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

SMILES string

O[C@H]([C@@H](OC(=O)\C=C\c1ccc(O)c(O)c1)C(O)=O)C(O)=O

InChI

1S/C13H12O9/c14-7-3-1-6(5-8(7)15)2-4-9(16)22-11(13(20)21)10(17)12(18)19/h1-5,10-11,14-15,17H,(H,18,19)(H,20,21)/b4-2+/t10-,11-/m1/s1

InChI key

SWGKAHCIOQPKFW-JTNORFRNSA-N

General description

Cafteric acid (CFA) is a polyphenolic compound belonging to the hydroxycinnamic acids (HCAs) subgroup. It is a derivative of caffeic acid. Cafteric acid is mainly sourced from Echinacea purpurea.

Application

Caftaric acid has been used:
  • as a standard antioxidant to determine the antioxidant potential (AOP) of red wine using 2,2-diphenyl-1-picrylhydrazyl (DPPH) assay
  • as an antioxidant together with sulfur dioxide (SO2) to measure the antioxidant potential of white wines using 2,2-diphenyl-1-picrylhydrazyl (DPPH) and Folin-Ciocalteu (FC) assays
  • to evaluate the myelopoietic effect on bone marrow of rats treated with various Echinacea purpurea extracts
  • to identify (poly)phenolic compounds in concord grape juice and their metabolites in human plasma and urine after juice consumption

Biochem/physiol Actions

Caftaric acid exerts antioxidant and anti-inflammatory effects against indomethacin-induced gastric ulcers in rats. It also displays antimutagenicity properties.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.


Still not finding the right product?

Explore all of our products under Caftaric acid


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library