Skip to Content
Merck
All Photos(1)

Key Documents

14202

Sigma-Aldrich

Diisopropyl-fluorophosphatase

≥30 U/mg

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

fermentation/recombinant

Quality Level

form

lyophilized solid

specific activity

≥30 U/mg

storage temp.

−20°C

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Unit Definition

One unit corresponds to the amount of enzyme which hydrolyzes 1 μmol diisopropyl fluorophosphate per minute at pH 8.1 and 22 °C

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jürgen Gäb et al.
Analytical and bioanalytical chemistry, 396(3), 1213-1221 (2009-11-28)
The enzyme diisopropyl fluorophosphatase (DFPase, EC 3.1.8.2) from the squid Loligo vulgaris effectively catalyzes the hydrolysis of diisopropyl fluorophosphate (DFP) and a number of organophosphorus nerve agents, including sarin, soman, cyclosarin, and tabun. Until now, determination of kinetic data has
Marc-Michael Blum et al.
Biochemistry, 47(18), 5216-5224 (2008-04-10)
A wide range of toxic organophosphorus pesticides and nerve agents is effectively hydrolyzed by the structurally related phosphotriesterase enzymes paraoxonase (PON1) from human plasma and diisopropyl fluorophosphatase (DFPase) from the squid Loligo vulgaris. Both enzymes have potential use as medical
Debra A Gearhart et al.
Toxicology and applied pharmacology, 218(1), 20-29 (2006-11-25)
Diisopropylfluorophosphate, originally developed as a chemical warfare agent, is structurally similar to nerve agents, and chlorpyrifos has extensive worldwide use as an agricultural pesticide. While inhibition of cholinesterases underlies the acute toxicity of these organophosphates, we previously reported impaired axonal
Julian C H Chen et al.
Acta crystallographica. Section D, Biological crystallography, 66(Pt 11), 1131-1138 (2010-11-03)
Diisopropyl fluorophosphatase (DFPase) is a calcium-dependent phosphotriesterase that acts on a variety of highly toxic organophosphorus compounds that act as inhibitors of acetylcholinesterase. The mechanism of DFPase has been probed using a variety of methods, including isotopic labelling, which demonstrated
Marc-Michael Blum et al.
Proceedings of the National Academy of Sciences of the United States of America, 106(3), 713-718 (2009-01-13)
Hydrogen atoms constitute about half of all atoms in proteins and play a critical role in enzyme mechanisms and macromolecular and solvent structure. Hydrogen atom positions can readily be determined by neutron diffraction, and as such, neutron diffraction is an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service