Skip to Content
Merck
All Photos(1)

Key Documents

11631

Sigma-Aldrich

D-Glucosamine 3-sulfate

≥98.0% (TLC)

Synonym(s):

GlcN-3S

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13NO8S
CAS Number:
Molecular Weight:
259.23
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥98.0% (TLC)

form

powder

optical activity

[α]/D 55.0±2.0

technique(s)

thin layer chromatography (TLC): suitable

storage temp.

−20°C

SMILES string

N[C@H]1C(O)O[C@H](CO)[C@@H](O)[C@@H]1OS(O)(=O)=O

InChI

1S/C6H13NO8S/c7-3-5(15-16(11,12)13)4(9)2(1-8)14-6(3)10/h2-6,8-10H,1,7H2,(H,11,12,13)/t2-,3-,4-,5-,6?/m1/s1

InChI key

UZUBNIPDAIVWIE-IVMDWMLBSA-N

Application

D-Glucosamine 3-sulfate (GlcN-3S) may be used as a reference in analytical analysis of the components of heparin sulfate.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A S Edge et al.
The Journal of biological chemistry, 265(26), 15874-15881 (1990-09-15)
Fragmentation of the heparan sulfate chains from bovine glomerular basement membrane (GBM) by hydrazine/nitrous acid treatment followed by NaB3H4-reduction yielded a mixture of six sulfated disaccharides containing D-glucuronic (GlcUA) or L-iduronic acid (IdUA) and terminating in 2,5-anhydro[3H]mannitol (AnManH2), in addition
H G Garg et al.
Biochemical and biophysical research communications, 224(2), 468-473 (1996-07-16)
Heparin macromolecules inhibit vascular remodeling associated with hypoxic pulmonary hypertension. Heparin's antiproliferative effect on smooth muscle cells, based on studies of synthetic pentasaccharide fragments, has been attributed to 3-O-sulfate on the internal glucosamine. To determine the role of 3-O-sulfation in
H Tsuda et al.
The Journal of biological chemistry, 271(18), 10495-10502 (1996-05-03)
Porcine intestinal heparin was extensively digested with Flavobacterium heparinase and size-fractionated by gel chromatography. Subfractionation of the hexasaccharide fraction by anion exchange high pressure liquid chromatography yielded 10 fractions. Six contained oligosaccharides derived from the repeating disaccharide region, whereas four
Alessandro Parra et al.
Glycobiology, 22(2), 248-257 (2011-09-22)
Glycosaminoglycans were extracted from both young rabbit growth plate (GRP) and articular (ART) cartilage tissues and enzymatically treated to selectively eliminate chondroitin sulfates and hyaluronic acid. The procedure avoided any fractionation step that could enrich the extract with over- or
A Naggi et al.
Carbohydrate research, 336(4), 283-290 (2001-12-01)
In the framework of a project aimed at generating heparin-like sulfation patterns and biological activities in biotechnological glycosaminoglycans, different approaches have been considered for simulating the alpha(1-->4)-linked 2-O-sulfated L-iduronic acid (IdoA2SO(3))-->N,6-O-sulfated D-glucosamine (GlcNSO(3)6SO(3)) disaccharide sequences prevalent in mammalian heparins. Since

Articles

Glycosaminoglycans are large linear polysaccharides constructed of repeating disaccharide units.

Glycosaminoglycans are large linear polysaccharides constructed of repeating disaccharide units.

Glycosaminoglycans are large linear polysaccharides constructed of repeating disaccharide units.

Glycosaminoglycans are large linear polysaccharides constructed of repeating disaccharide units.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service