Recommended Products
biological source
human esophagus
Quality Level
growth mode
Adherent
karyotype
Modal number 58 chromosomal aberration at 11p13-15.
morphology
Epithelial
products
Expression of actin imtermediate filaments and growth factors TGF?1, TGF?2, TGF?3, TGF? and platelet-derived growth factor A (PDGFA).
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
metastasis
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Cell Line Description
DNA Profile
CSF1PO: 10
D13S317: 12,13
D16S539: 11,12
D5S818: 10
D7S820: 9,10
THO1: 8,9
TPOX: 9,12
vWA: 15,18
Culture Medium
Subculture Routine
Other Notes
Disclaimer
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service