Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C11H19NO3
CAS Number:
Molecular Weight:
213.27
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26
MDL number:
Product Name
N-Heptanoyl-DL-homoserine lactone, ≥97.0% (HPLC)
assay
≥97.0% (HPLC)
form
powder with small lumps
color
white to faintly brown
storage temp.
−20°C
SMILES string
CCCCCCC(=O)NC1CCOC1=O
InChI
1S/C11H19NO3/c1-2-3-4-5-6-10(13)12-9-7-8-15-11(9)14/h9H,2-8H2,1H3,(H,12,13)
InChI key
FTMZLSDESAOPSZ-UHFFFAOYSA-N
Application
Application test: Induces violacein expression in a Chromobacterium violaceum mutant usually not able to produce homoserine lactones.
Biochem/physiol Actions
N-Heptanoyl-DL-homoserine lactone (C7HSL) is among a group of homoserine lactones that includes; N-octanoyl-homoserine lactone (N-C8-HSL), N-Decanoyl-DL-homoserine lactone (N-C10-HSL), N-(3-oxodecanoyl) homoserine-L-lactone (3-oxo-C10 HSL), N-(3-oxododecanoyl)homoserine-L-lactone (3-oxo-C12-HSL), N-(3-Oxotetradecanoyl)-L-homoserine lactone (3-oxo-C14-HSL, N-(3-hydroxydecanoyl)-L-homoserine lactone, and N-(3-hydroxyoctanoyl)-L-homoserine lactone involved in the processes of bacterial quorum sensing. These N-acyl-homoserine lactones are used to study the processes and mechanisms of bacterial quorum sensing.
N-Heptanoyl-DL-homoserine lactone is a member of N-acyl-homoserine lactone family. N-acylhomoserine lactones (AHL) regulate gene expression in gram-negative bacteria, such asEcherichia and Salmonella are involved in quorum sensing, cell to cell communication among bacteria. Some AHLs are potent chemoattractants for human immune cells such as neutrophils.
Packaging
Bottomless glass bottle. Contents are inside inserted fused cone.
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 944076-1KG | 04065273306149 |
| 203114-5G | 04061838765635 |
| 221643-50G | 04061838252555 |
| 221643-250G | 04061838777454 |
| 637025-100G | 04061832723419 |
| 637025-25G | 04061832723433 |
| 637025-250G | 04061832723426 |
| 944076-100G | 04065273306132 |
| 10939-25MG | 04061838688385 |