Skip to Content
Merck

10080

α-Amylase from hog pancreas

powder, ~50 U/mg

Synonym(s):

α-Amylase Enzyme

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352204
EC Number:
232-565-6
NACRES:
NA.54
Specific activity:
~50 U/mg
Biological source:
hog pancreas
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

hog pancreas

Quality Level

form

powder

specific activity

~50 U/mg

storage temp.

−20°C

General description

Pancreatic α-amylase is produced by the pancreatic acinar cells and released into the duodenum. It is a major molecule of pancreatic fluid.

Application

α-Amylase from hog pancreas has been used:
  • in the in vitro digestion system to study the hydrolysis of pea globulins
  • in vitro α-amylase inhibition study
  • to prepare digestive juice (salivary juice) for the in vitro model of the human digestion system

Biochem/physiol Actions

α-Amylase is a starch hydrolase, for glucose production, important for energy acquisition. It catalyzes the hydrolysis of α-1,4-glycosidic linkages in starch to form glucose, maltose, and maltotriose units. In mammals, α-amylase regulates the sugar assimilation by reducing the glucose uptake by Na+/glucose cotransporter 1 (SGLT1) and stimulating glycoprotein N-glycans mediated starch digestion. α-Amylase inhibitors are associated with the treatment of type 2 diabetes mellitus. α-Amylases are widely known industrial enzymes used in food, detergent, textile, fermentation, and pharmaceutical industries.

Other Notes

1 U corresponds to the amount of enzyme which liberates 1 μmol maltose per minute at pH 6.9 and 25°C (starch acc. to Zulkowsky, Cat. No. 85642, as substrate)
Optimum pH and temperature 6.9 and 53 °C, respectively; calcium is required for activity and also prevents degradation by trypsin; Characterization.


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Paula Monteiro de Souza et al.
Brazilian journal of microbiology : [publication of the Brazilian Society for Microbiology], 41(4), 850-861 (2010-10-01)
Amylases are one of the main enzymes used in industry. Such enzymes hydrolyze the starch molecules into polymers composed of glucose units. Amylases have potential application in a wide number of industrial processes such as food, fermentation and pharmaceutical industries.
Muhammad H Alu'datt et al.
Food chemistry, 240, 784-798 (2017-09-28)
This investigation was performed to assess the effects of sonication on the structure of protein, extractability of phenolics, and biological properties of isolated proteins and protein co-precipitates prepared from brewers' spent grain and soybean flour. Scanning electron micrographs revealed that
Yanmei Huang et al.
Food chemistry, 344, 128610-128610 (2020-11-23)
This study was designed to investigate quantitatively the inhibition and molecular mechanism of pancreatic α-amylase exhibited by flavonoids from dandelion to reveal its potential use in relieving postprandial hyperglycemia. The results show that the flavonoids reversibly inhibited the α-amylase in



Global Trade Item Number

SKUGTIN
10080-25G04061838664273
10080-100G04061838664266